The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001394.6 | NP_001385.1 | 1 - 2 | 904 | 62 |
GenBank mRNA | AK314820.1 | - | 1 - 2 | 829 | 62 |
BC002671.2 | - | 1 - 2 | 590 | 62 | |
BC014565.2 | - | 1 - 2 | 822 | 62 | |
S80632.1 | - | 1 - 2 | 483 | 62 | |
U21108.1 | - | 1 - 2 | 678 | 62 | |
U48807.1 | - | 1 - 2 | 551 | 62 |