The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243080.1 | NP_001230009.1 | 1 - 2 | 122 | 85 |
NM_001422.3 | NP_001413.1 | 1 - 2 | 122 | 85 | |
XM_017017308.1 | XP_016872797.1 | 1 - 2 | 87 | 85 | |
XM_017017309.1 | XP_016872798.1 | 1 - 2 | 87 | 85 | |
GenBank mRNA | AF049703.1 | - | 1 - 2 | 103 | 85 |
AF115403.1 | - | 1 - 2 | 90 | 85 | |
AK074633.1 | - | 1 - 2 | 88 | 85 | |
AK075197.1 | - | 1 - 2 | 78 | 85 | |
AK290808.1 | - | 1 - 2 | 78 | 85 | |
AK312537.1 | - | 1 - 2 | 116 | 85 | |
BC029743.1 | - | 1 - 2 | 105 | 85 | |
BG260616.1 | - | - | 105 | 85 | |
BP358543.1 | - | 1 - 2 | 122 | 85 | |
DQ123839.1 | - | 1 - 2 | 127 | 85 |