Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAT2
Entrez Gene ID
2196
Gene Name
FAT atypical cadherin 2
Gene Aliases
CDHF8, CDHR9, HFAT2, MEGF1
Chromosome Location
Chr.5: 151504092 - 151594822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001447.2 NP_001438.1 12 - 13 9438 76
XM_006714761.3 XP_006714824.1 13 - 14 10009 76
XM_011537600.2 XP_011535902.1 15 - 16 9777 76
XM_011537603.2 XP_011535905.1 13 - 14 9710 76
XM_017009224.1 XP_016864713.1 15 - 16 9779 76
XM_017009225.1 XP_016864714.1 14 - 15 9621 76
GenBank mRNA AF231022.1 - 12 - 13 9438 76

View Full Product Details