Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31357012-31357533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGAX
Entrez Gene ID
3687
Gene Name
integrin subunit alpha X
Gene Aliases
CD11C, SLEB6
UniGene
Hs.248472
Gene Chromosome Location
Chr.16: 31355134-31382997 on Build GRCh38
Transcripts
NM_000887.4 NM_001286375.1 XM_011545852.1 XM_011545854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4491680 c.384C>T p.L128L Chr.16: 31357318-31357318 on Build GRCh38
3402301 c.333C>T p.T111T Chr.16: 31357267-31357267 on Build GRCh38
4849485 c.414C>T p.L138L Chr.16: 31357348-31357348 on Build GRCh38
3772202 c.248-9T>A p.? Chr.16: 31357022-31357022 on Build GRCh38
4060442 c.342C>T p.H114H Chr.16: 31357276-31357276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558222544 Chr.16: 31357303-31357303 on Build GRCh38
rs780063353 Chr.16: 31357401-31357401 on Build GRCh38
rs753233568 Chr.16: 31357096-31357096 on Build GRCh38
rs373622931 Chr.16: 31357045-31357045 on Build GRCh38
rs781611809 Chr.16: 31357037-31357037 on Build GRCh38
rs771874041 Chr.16: 31357160-31357160 on Build GRCh38
rs780480088 Chr.16: 31357113-31357113 on Build GRCh38
rs370805911 Chr.16: 31357043-31357043 on Build GRCh38
rs374342177 Chr.16: 31357214-31357214 on Build GRCh38
rs201858672 Chr.16: 31357394-31357394 on Build GRCh38
rs745409254 Chr.16: 31357132-31357135 on Build GRCh38
rs369137615 Chr.16: 31357300-31357300 on Build GRCh38
rs11574636 Chr.16: 31357514-31357514 on Build GRCh38
rs755431431 Chr.16: 31357120-31357120 on Build GRCh38
rs761480055 Chr.16: 31357267-31357267 on Build GRCh38
rs756783395 Chr.16: 31357097-31357097 on Build GRCh38
rs780864207 Chr.16: 31357406-31357406 on Build GRCh38
rs769431411 Chr.16: 31357318-31357318 on Build GRCh38
rs761272220 Chr.16: 31357195-31357195 on Build GRCh38
rs750617140 Chr.16: 31357393-31357393 on Build GRCh38
rs779971705 Chr.16: 31357247-31357247 on Build GRCh38
rs545813521 Chr.16: 31357194-31357194 on Build GRCh38
rs768267505 Chr.16: 31357454-31357454 on Build GRCh38
rs377514607 Chr.16: 31357151-31357151 on Build GRCh38
rs747761782 Chr.16: 31357315-31357315 on Build GRCh38
rs752081198 Chr.16: 31357115-31357115 on Build GRCh38
rs61761295 Chr.16: 31357258-31357258 on Build GRCh38
rs749493938 Chr.16: 31357233-31357233 on Build GRCh38
rs757328876 Chr.16: 31357237-31357237 on Build GRCh38
rs141436259 Chr.16: 31357301-31357301 on Build GRCh38
rs371786173 Chr.16: 31357074-31357074 on Build GRCh38
rs745832842 Chr.16: 31357240-31357240 on Build GRCh38
rs771925567 Chr.16: 31357248-31357248 on Build GRCh38
rs778999205 Chr.16: 31357238-31357238 on Build GRCh38
rs1004906993 Chr.16: 31357467-31357467 on Build GRCh38
rs754027414 Chr.16: 31357291-31357291 on Build GRCh38
rs771429171 Chr.16: 31357160-31357160 on Build GRCh38
rs768196583 Chr.16: 31357272-31357272 on Build GRCh38
rs531724454 Chr.16: 31357412-31357412 on Build GRCh38
rs761950784 Chr.16: 31357373-31357373 on Build GRCh38
rs756474155 Chr.16: 31357282-31357282 on Build GRCh38
rs868016039 Chr.16: 31357486-31357486 on Build GRCh38
rs267604519 Chr.16: 31357263-31357263 on Build GRCh38
rs889567157 Chr.16: 31357507-31357507 on Build GRCh38
rs774193687 Chr.16: 31357157-31357157 on Build GRCh38
rs572322134 Chr.16: 31357159-31357159 on Build GRCh38
rs759445520 Chr.16: 31357158-31357158 on Build GRCh38
rs201621258 Chr.16: 31357322-31357322 on Build GRCh38
rs748402544 Chr.16: 31357147-31357147 on Build GRCh38
rs754880697 Chr.16: 31357405-31357405 on Build GRCh38
rs748347429 Chr.16: 31357038-31357038 on Build GRCh38
rs769126054 Chr.16: 31357033-31357033 on Build GRCh38
rs763619758 Chr.16: 31357092-31357092 on Build GRCh38
rs371278987 Chr.16: 31357506-31357506 on Build GRCh38
rs762618934 Chr.16: 31357339-31357339 on Build GRCh38
rs760283854 Chr.16: 31357080-31357080 on Build GRCh38
rs762961702 Chr.16: 31357044-31357044 on Build GRCh38
rs760320633 Chr.16: 31357180-31357180 on Build GRCh38
rs61761294 Chr.16: 31357184-31357184 on Build GRCh38
rs11574635 Chr.16: 31357495-31357495 on Build GRCh38
rs748033148 Chr.16: 31357411-31357411 on Build GRCh38
rs763716541 Chr.16: 31357185-31357185 on Build GRCh38
rs924610893 Chr.16: 31357136-31357136 on Build GRCh38
rs770533015 Chr.16: 31357350-31357350 on Build GRCh38
rs764495770 Chr.16: 31357287-31357287 on Build GRCh38
rs754161938 Chr.16: 31357369-31357369 on Build GRCh38
rs765497772 Chr.16: 31357386-31357386 on Build GRCh38
rs746049384 Chr.16: 31357165-31357165 on Build GRCh38
rs150827519 Chr.16: 31357304-31357304 on Build GRCh38
rs771037325 Chr.16: 31357155-31357155 on Build GRCh38
rs775273310 Chr.16: 31357249-31357249 on Build GRCh38
rs941623746 Chr.16: 31357472-31357473 on Build GRCh38
rs778859699 Chr.16: 31357302-31357302 on Build GRCh38
rs761193825 Chr.16: 31357351-31357351 on Build GRCh38
rs767998541 Chr.16: 31357222-31357222 on Build GRCh38
rs148375692 Chr.16: 31357051-31357051 on Build GRCh38
rs370901437 Chr.16: 31357229-31357229 on Build GRCh38
rs564218640 Chr.16: 31357276-31357276 on Build GRCh38
rs770023120 Chr.16: 31357042-31357042 on Build GRCh38
rs775221870 Chr.16: 31357175-31357175 on Build GRCh38
rs912208552 Chr.16: 31357292-31357292 on Build GRCh38
rs753053210 Chr.16: 31357227-31357227 on Build GRCh38
rs776448408 Chr.16: 31357306-31357306 on Build GRCh38
rs764396019 Chr.16: 31357360-31357360 on Build GRCh38
rs374769572 Chr.16: 31357280-31357280 on Build GRCh38
rs1015157912 Chr.16: 31357199-31357199 on Build GRCh38
rs781654631 Chr.16: 31357126-31357126 on Build GRCh38
rs746876765 Chr.16: 31357250-31357250 on Build GRCh38
rs764785024 Chr.16: 31357200-31357200 on Build GRCh38
rs373801738 Chr.16: 31357402-31357402 on Build GRCh38
rs760133637 Chr.16: 31357268-31357268 on Build GRCh38
rs1029116658 Chr.16: 31357515-31357515 on Build GRCh38
rs906511509 Chr.16: 31357448-31357448 on Build GRCh38
rs575067881 Chr.16: 31357098-31357098 on Build GRCh38
rs973778392 Chr.16: 31357040-31357040 on Build GRCh38
rs998263838 Chr.16: 31357503-31357503 on Build GRCh38
rs973557610 Chr.16: 31357245-31357245 on Build GRCh38
rs747016909 Chr.16: 31357363-31357363 on Build GRCh38
rs554450360 Chr.16: 31357152-31357152 on Build GRCh38
rs372129439 Chr.16: 31357217-31357217 on Build GRCh38
rs776198821 Chr.16: 31357259-31357259 on Build GRCh38

View Full Product Details