Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31359610-31360118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGAX
Entrez Gene ID
3687
Gene Name
integrin subunit alpha X
Gene Aliases
CD11C, SLEB6
UniGene
Hs.248472
Gene Chromosome Location
Chr.16: 31355134-31382997 on Build GRCh38
Transcripts
NM_000887.4 NM_001286375.1 XM_011545852.1 XM_011545854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4060444 c.691G>A p.A231T Chr.16: 31360049-31360049 on Build GRCh38
3787145 c.485T>C p.I162T Chr.16: 31359754-31359754 on Build GRCh38
3387406 c.507G>A p.T169T Chr.16: 31359776-31359776 on Build GRCh38
2917963 c.705C>T p.V235V Chr.16: 31360063-31360063 on Build GRCh38
4060443 c.519C>T p.F173F Chr.16: 31359788-31359788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754482275 Chr.16: 31359934-31359934 on Build GRCh38
rs750130802 Chr.16: 31360027-31360027 on Build GRCh38
rs751601296 Chr.16: 31359694-31359694 on Build GRCh38
rs367628555 Chr.16: 31359858-31359858 on Build GRCh38
rs760900105 Chr.16: 31359994-31359994 on Build GRCh38
rs543041070 Chr.16: 31359645-31359645 on Build GRCh38
rs139287488 Chr.16: 31359767-31359767 on Build GRCh38
rs777608495 Chr.16: 31359649-31359649 on Build GRCh38
rs374289179 Chr.16: 31359797-31359797 on Build GRCh38
rs773956136 Chr.16: 31359662-31359662 on Build GRCh38
rs781257815 Chr.16: 31359703-31359703 on Build GRCh38
rs906812422 Chr.16: 31359686-31359686 on Build GRCh38
rs146081281 Chr.16: 31359775-31359775 on Build GRCh38
rs757924387 Chr.16: 31359922-31359922 on Build GRCh38
rs768821650 Chr.16: 31359995-31359995 on Build GRCh38
rs199871999 Chr.16: 31359762-31359762 on Build GRCh38
rs1018777948 Chr.16: 31359826-31359826 on Build GRCh38
rs746580487 Chr.16: 31359933-31359933 on Build GRCh38
rs774736704 Chr.16: 31359981-31359981 on Build GRCh38
rs368541545 Chr.16: 31359917-31359917 on Build GRCh38
rs773697965 Chr.16: 31359844-31359844 on Build GRCh38
rs115153713 Chr.16: 31359990-31359990 on Build GRCh38
rs376273425 Chr.16: 31360060-31360060 on Build GRCh38
rs765087677 Chr.16: 31360019-31360019 on Build GRCh38
rs770639010 Chr.16: 31359743-31359743 on Build GRCh38
rs774432855 Chr.16: 31359778-31359778 on Build GRCh38
rs779347658 Chr.16: 31360058-31360058 on Build GRCh38
rs202064325 Chr.16: 31359845-31359845 on Build GRCh38
rs964527732 Chr.16: 31360061-31360061 on Build GRCh38
rs767665066 Chr.16: 31359863-31359863 on Build GRCh38
rs748978672 Chr.16: 31359742-31359742 on Build GRCh38
rs751181508 Chr.16: 31360039-31360039 on Build GRCh38
rs1047037565 Chr.16: 31359864-31359864 on Build GRCh38
rs1043448553 Chr.16: 31359980-31359980 on Build GRCh38
rs761088077 Chr.16: 31359677-31359677 on Build GRCh38
rs991851439 Chr.16: 31359709-31359709 on Build GRCh38
rs780459216 Chr.16: 31360062-31360062 on Build GRCh38
rs375368829 Chr.16: 31360064-31360064 on Build GRCh38
rs750669635 Chr.16: 31359683-31359683 on Build GRCh38
rs4264407 Chr.16: 31359872-31359872 on Build GRCh38
rs201048460 Chr.16: 31360041-31360041 on Build GRCh38
rs769145495 Chr.16: 31359679-31359679 on Build GRCh38
rs778576565 Chr.16: 31359745-31359745 on Build GRCh38
rs773198238 Chr.16: 31360070-31360070 on Build GRCh38
rs763982466 Chr.16: 31359824-31359824 on Build GRCh38
rs753626983 Chr.16: 31359828-31359828 on Build GRCh38
rs757953328 Chr.16: 31359839-31359839 on Build GRCh38
rs200189050 Chr.16: 31359833-31359833 on Build GRCh38
rs1055396594 Chr.16: 31359930-31359930 on Build GRCh38
rs371816178 Chr.16: 31359868-31359868 on Build GRCh38
rs745489546 Chr.16: 31359838-31359838 on Build GRCh38
rs149925845 Chr.16: 31359992-31359992 on Build GRCh38
rs762698318 Chr.16: 31360072-31360072 on Build GRCh38
rs752546467 Chr.16: 31359724-31359724 on Build GRCh38
rs145143041 Chr.16: 31360036-31360036 on Build GRCh38
rs757155175 Chr.16: 31359903-31359903 on Build GRCh38
rs200320931 Chr.16: 31360049-31360049 on Build GRCh38
rs745437742 Chr.16: 31359749-31359749 on Build GRCh38
rs759700394 Chr.16: 31359788-31359788 on Build GRCh38
rs771474273 Chr.16: 31359758-31359758 on Build GRCh38
rs372032821 Chr.16: 31360063-31360063 on Build GRCh38
rs776735511 Chr.16: 31360000-31360000 on Build GRCh38
rs191713688 Chr.16: 31359842-31359842 on Build GRCh38
rs763123555 Chr.16: 31359685-31359685 on Build GRCh38
rs891895732 Chr.16: 31359873-31359873 on Build GRCh38
rs370729027 Chr.16: 31359795-31359795 on Build GRCh38
rs376541940 Chr.16: 31360030-31360030 on Build GRCh38
rs748925673 Chr.16: 31359651-31359651 on Build GRCh38
rs754999681 Chr.16: 31359695-31359695 on Build GRCh38
rs759627349 Chr.16: 31359860-31359860 on Build GRCh38
rs1574566 Chr.16: 31359961-31359961 on Build GRCh38
rs767775627 Chr.16: 31360066-31360066 on Build GRCh38
rs764896332 Chr.16: 31359909-31359909 on Build GRCh38
rs552168067 Chr.16: 31359794-31359794 on Build GRCh38
rs749773577 Chr.16: 31359958-31359958 on Build GRCh38
rs892527127 Chr.16: 31359634-31359634 on Build GRCh38
rs766453540 Chr.16: 31359693-31359693 on Build GRCh38
rs913982168 Chr.16: 31359858-31359858 on Build GRCh38
rs748559101 Chr.16: 31359763-31359763 on Build GRCh38
rs764182652 Chr.16: 31359891-31359891 on Build GRCh38
rs75652952 Chr.16: 31360048-31360048 on Build GRCh38
rs753818450 Chr.16: 31359896-31359896 on Build GRCh38
rs549906598 Chr.16: 31359726-31359726 on Build GRCh38
rs960225230 Chr.16: 31359688-31359688 on Build GRCh38
rs372254909 Chr.16: 31359955-31359955 on Build GRCh38
rs779632742 Chr.16: 31359926-31359926 on Build GRCh38
rs938271101 Chr.16: 31359916-31359916 on Build GRCh38
rs770585827 Chr.16: 31359658-31359658 on Build GRCh38
rs746105917 Chr.16: 31359987-31359987 on Build GRCh38
rs376523474 Chr.16: 31359789-31359789 on Build GRCh38
rs771349695 Chr.16: 31359967-31359967 on Build GRCh38
rs373380635 Chr.16: 31359735-31359735 on Build GRCh38
rs139466358 Chr.16: 31359728-31359728 on Build GRCh38
rs779393714 Chr.16: 31359841-31359841 on Build GRCh38
rs45509092 Chr.16: 31359847-31359847 on Build GRCh38
rs777661708 Chr.16: 31359737-31359737 on Build GRCh38

View Full Product Details