Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31361971-31362476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGAX
Entrez Gene ID
3687
Gene Name
integrin subunit alpha X
Gene Aliases
CD11C, SLEB6
UniGene
Hs.248472
Gene Chromosome Location
Chr.16: 31355134-31382997 on Build GRCh38
Transcripts
NM_000887.4 NM_001286375.1 XM_011545852.1 XM_011545854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4910432 c.1171A>T p.I391F Chr.16: 31362159-31362159 on Build GRCh38
3988448 c.1087-1G>A p.? Chr.16: 31362074-31362074 on Build GRCh38
703163 c.1105G>T p.A369S Chr.16: 31362093-31362093 on Build GRCh38
4521802 c.1130G>A p.G377E Chr.16: 31362118-31362118 on Build GRCh38
970208 c.1141C>A p.L381M Chr.16: 31362129-31362129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377459367 Chr.16: 31362253-31362253 on Build GRCh38
rs766485722 Chr.16: 31362055-31362055 on Build GRCh38
rs759649296 Chr.16: 31362074-31362074 on Build GRCh38
rs748609911 Chr.16: 31362116-31362116 on Build GRCh38
rs759573834 Chr.16: 31362143-31362143 on Build GRCh38
rs200606612 Chr.16: 31362112-31362112 on Build GRCh38
rs778496418 Chr.16: 31362096-31362096 on Build GRCh38
rs760646360 Chr.16: 31362159-31362159 on Build GRCh38
rs371693236 Chr.16: 31362118-31362118 on Build GRCh38
rs779503013 Chr.16: 31362214-31362214 on Build GRCh38
rs895230426 Chr.16: 31362372-31362372 on Build GRCh38
rs765697666 Chr.16: 31362401-31362401 on Build GRCh38
rs756790585 Chr.16: 31362094-31362094 on Build GRCh38
rs997324002 Chr.16: 31361995-31361995 on Build GRCh38
rs777934960 Chr.16: 31362128-31362128 on Build GRCh38
rs961620669 Chr.16: 31362283-31362283 on Build GRCh38
rs749971517 Chr.16: 31362101-31362101 on Build GRCh38
rs970686177 Chr.16: 31362424-31362424 on Build GRCh38
rs750070858 Chr.16: 31362196-31362196 on Build GRCh38
rs889161682 Chr.16: 31362097-31362098 on Build GRCh38
rs747552846 Chr.16: 31362031-31362031 on Build GRCh38
rs202066072 Chr.16: 31362179-31362179 on Build GRCh38
rs775703951 Chr.16: 31362134-31362134 on Build GRCh38
rs367918770 Chr.16: 31362109-31362109 on Build GRCh38
rs959799331 Chr.16: 31361998-31361998 on Build GRCh38
rs772309162 Chr.16: 31362240-31362240 on Build GRCh38
rs891107627 Chr.16: 31362001-31362001 on Build GRCh38
rs976710228 Chr.16: 31362300-31362300 on Build GRCh38
rs200873409 Chr.16: 31362047-31362047 on Build GRCh38
rs983930978 Chr.16: 31362022-31362022 on Build GRCh38
rs200403295 Chr.16: 31362062-31362062 on Build GRCh38
rs375007025 Chr.16: 31362141-31362141 on Build GRCh38
rs745537840 Chr.16: 31362230-31362230 on Build GRCh38
rs753554504 Chr.16: 31362093-31362093 on Build GRCh38
rs201470398 Chr.16: 31362009-31362009 on Build GRCh38
rs748554785 Chr.16: 31362040-31362040 on Build GRCh38
rs1012279494 Chr.16: 31362396-31362396 on Build GRCh38
rs760698150 Chr.16: 31362244-31362244 on Build GRCh38
rs768984349 Chr.16: 31362035-31362035 on Build GRCh38
rs138866008 Chr.16: 31362175-31362175 on Build GRCh38
rs767634085 Chr.16: 31362089-31362089 on Build GRCh38
rs759594937 Chr.16: 31362084-31362084 on Build GRCh38
rs757876377 Chr.16: 31362105-31362105 on Build GRCh38
rs778330950 Chr.16: 31362220-31362220 on Build GRCh38
rs184225456 Chr.16: 31362225-31362225 on Build GRCh38
rs397967107 Chr.16: 31362363-31362364 on Build GRCh38
rs577643926 Chr.16: 31362123-31362123 on Build GRCh38
rs773448780 Chr.16: 31362044-31362044 on Build GRCh38
rs1015423670 Chr.16: 31362272-31362272 on Build GRCh38
rs202053012 Chr.16: 31362042-31362042 on Build GRCh38
rs370314716 Chr.16: 31362060-31362060 on Build GRCh38
rs746038387 Chr.16: 31362237-31362237 on Build GRCh38
rs116039611 Chr.16: 31362266-31362266 on Build GRCh38
rs922499257 Chr.16: 31362318-31362318 on Build GRCh38
rs1008244665 Chr.16: 31362002-31362002 on Build GRCh38
rs767627747 Chr.16: 31362083-31362083 on Build GRCh38
rs775591508 Chr.16: 31362158-31362158 on Build GRCh38
rs530118018 Chr.16: 31362191-31362191 on Build GRCh38
rs771294576 Chr.16: 31362226-31362226 on Build GRCh38
rs772255897 Chr.16: 31362148-31362148 on Build GRCh38
rs554656941 Chr.16: 31362039-31362039 on Build GRCh38
rs374293554 Chr.16: 31362092-31362092 on Build GRCh38
rs200734472 Chr.16: 31362243-31362243 on Build GRCh38
rs753207706 Chr.16: 31362216-31362216 on Build GRCh38
rs138501712 Chr.16: 31362136-31362136 on Build GRCh38
rs529301620 Chr.16: 31362015-31362015 on Build GRCh38
rs375590428 Chr.16: 31362134-31362134 on Build GRCh38
rs60020095 Chr.16: 31362361-31362362 on Build GRCh38
rs756556451 Chr.16: 31362219-31362219 on Build GRCh38
rs1028434352 Chr.16: 31361996-31361996 on Build GRCh38
rs546424845 Chr.16: 31362323-31362323 on Build GRCh38
rs757929520 Chr.16: 31362204-31362204 on Build GRCh38
rs74015512 Chr.16: 31362012-31362012 on Build GRCh38

View Full Product Details