The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321015.1 | NP_001307944.1 | 5 - 6 | 931 | 122 |
NM_001482.2 | NP_001473.1 | 2 - 3 | 623 | 122 | |
GenBank mRNA | AK098055.1 | - | 1 - 2 | 1950 | 122 |
AK098393.1 | - | 2 - 3 | 425 | 122 | |
AK223585.1 | - | 2 - 3 | 385 | 122 | |
AK294995.1 | - | 2 - 3 | 389 | 122 | |
BC004141.2 | - | 2 - 3 | 364 | 122 | |
S68805.1 | - | 2 - 3 | 363 | 122 |