Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GBAS
Entrez Gene ID
2631
Gene Name
glioblastoma amplified sequence
Gene Aliases
NIPSNAP2
Chromosome Location
Chr.7: 55964577 - 56000182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001483.2 NP_001474.1 3 - 4 313 64
GenBank mRNA AF029786.1 - 3 - 4 288 64
AJ001259.1 - 3 - 4 280 64
AK315648.1 - 3 - 4 298 64
BC000732.2 - 3 - 4 286 64
BC001837.2 - 3 - 4 286 64
BC030821.1 - 3 - 4 295 64
BT007112.1 - 3 - 4 280 64
CR407612.1 - 3 - 4 280 64
CX166452.1 - 3 - 4 313 64

View Full Product Details