Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46659561-46660073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS35
Entrez Gene ID
55737
Gene Name
VPS35 retromer complex component
Gene Aliases
MEM3, PARK17
UniGene
Hs.454528
Gene Chromosome Location
Chr.16: 46657979-46689232 on Build GRCh38
Transcripts
NM_018206.5 XM_005256045.3

SNP Detail

SNP ID Chromosome Location
rs1006755372 Chr.16: 46659724-46659724 on Build GRCh38
rs774565598 Chr.16: 46659971-46659971 on Build GRCh38
rs1012263955 Chr.16: 46659789-46659789 on Build GRCh38
rs968372667 Chr.16: 46660019-46660019 on Build GRCh38
rs985863143 Chr.16: 46659959-46659959 on Build GRCh38
rs976022796 Chr.16: 46659746-46659746 on Build GRCh38
rs183646374 Chr.16: 46659708-46659708 on Build GRCh38
rs894312500 Chr.16: 46659773-46659773 on Build GRCh38
rs779756007 Chr.16: 46659847-46659847 on Build GRCh38
rs1032102320 Chr.16: 46659767-46659767 on Build GRCh38
rs1045140300 Chr.16: 46659803-46659803 on Build GRCh38
rs948904221 Chr.16: 46659864-46659864 on Build GRCh38
rs938523361 Chr.16: 46659710-46659710 on Build GRCh38
rs754785560 Chr.16: 46659885-46659885 on Build GRCh38
rs560695968 Chr.16: 46659591-46659591 on Build GRCh38
rs1021409720 Chr.16: 46660006-46660006 on Build GRCh38
rs749705519 Chr.16: 46659828-46659828 on Build GRCh38
rs191441368 Chr.16: 46659646-46659646 on Build GRCh38
rs953537707 Chr.16: 46659936-46659936 on Build GRCh38
rs188743379 Chr.16: 46660018-46660018 on Build GRCh38
rs1698957 Chr.16: 46659628-46659628 on Build GRCh38
rs938925484 Chr.16: 46659956-46659956 on Build GRCh38
rs181198454 Chr.16: 46660050-46660050 on Build GRCh38
rs924564635 Chr.16: 46659684-46659684 on Build GRCh38
rs575736800 Chr.16: 46659658-46659658 on Build GRCh38
rs542974709 Chr.16: 46659889-46659889 on Build GRCh38
rs887760536 Chr.16: 46659813-46659813 on Build GRCh38
rs746602580 Chr.16: 46659855-46659855 on Build GRCh38
rs768759859 Chr.16: 46659897-46659897 on Build GRCh38
rs34926621 Chr.16: 46659710-46659711 on Build GRCh38
rs758047233 Chr.16: 46659733-46659733 on Build GRCh38
rs780263080 Chr.16: 46659612-46659612 on Build GRCh38
rs977527968 Chr.16: 46660030-46660030 on Build GRCh38
rs780922363 Chr.16: 46659888-46659888 on Build GRCh38
rs762727734 Chr.16: 46659860-46659860 on Build GRCh38
rs965268731 Chr.16: 46659735-46659735 on Build GRCh38
rs11550460 Chr.16: 46659772-46659772 on Build GRCh38
rs889649099 Chr.16: 46659710-46659710 on Build GRCh38
rs928836241 Chr.16: 46659906-46659906 on Build GRCh38
rs544973710 Chr.16: 46659709-46659710 on Build GRCh38

View Full Product Details