Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46674625-46675135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS35
Entrez Gene ID
55737
Gene Name
VPS35 retromer complex component
Gene Aliases
MEM3, PARK17
UniGene
Hs.454528
Gene Chromosome Location
Chr.16: 46657979-46689232 on Build GRCh38
Transcripts
NM_018206.5 XM_005256045.3 XM_011523227.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5828387 c.915-6_915-3delTTTT p.? Chr.16: 46674663-46674666 on Build GRCh38
3932356 c.948T>A p.P316P Chr.16: 46674627-46674627 on Build GRCh38
7309317 c.915-3_915-2insT p.? Chr.16: 46674662-46674663 on Build GRCh38
1609339 c.915-3delT p.? Chr.16: 46674663-46674663 on Build GRCh38
6400987 c.918A>T p.L306F Chr.16: 46674657-46674657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149807259 Chr.16: 46674882-46674882 on Build GRCh38
rs923959739 Chr.16: 46674922-46674922 on Build GRCh38
rs536134821 Chr.16: 46674830-46674830 on Build GRCh38
rs772965052 Chr.16: 46674663-46674663 on Build GRCh38
rs548079305 Chr.16: 46674780-46674780 on Build GRCh38
rs961970930 Chr.16: 46674723-46674723 on Build GRCh38
rs896649225 Chr.16: 46675090-46675090 on Build GRCh38
rs753610015 Chr.16: 46674675-46674675 on Build GRCh38
rs946815922 Chr.16: 46674839-46674839 on Build GRCh38
rs774933100 Chr.16: 46675056-46675056 on Build GRCh38
rs533745471 Chr.16: 46675055-46675055 on Build GRCh38
rs77133510 Chr.16: 46674660-46674660 on Build GRCh38
rs111447053 Chr.16: 46675013-46675013 on Build GRCh38
rs5816559 Chr.16: 46675010-46675011 on Build GRCh38
rs757976341 Chr.16: 46674678-46674678 on Build GRCh38
rs953255083 Chr.16: 46674780-46674781 on Build GRCh38
rs773736342 Chr.16: 46674711-46674711 on Build GRCh38
rs745511646 Chr.16: 46674677-46674677 on Build GRCh38
rs985929003 Chr.16: 46674785-46674789 on Build GRCh38
rs796765129 Chr.16: 46674663-46674664 on Build GRCh38
rs757051230 Chr.16: 46674676-46674676 on Build GRCh38
rs1057151447 Chr.16: 46675019-46675019 on Build GRCh38
rs377192015 Chr.16: 46674679-46674679 on Build GRCh38
rs374327750 Chr.16: 46674726-46674726 on Build GRCh38
rs569369937 Chr.16: 46674663-46674664 on Build GRCh38
rs776216302 Chr.16: 46674663-46674665 on Build GRCh38
rs140065133 Chr.16: 46674740-46674740 on Build GRCh38
rs756137934 Chr.16: 46674657-46674657 on Build GRCh38
rs868269687 Chr.16: 46674655-46674655 on Build GRCh38
rs547075243 Chr.16: 46674682-46674682 on Build GRCh38
rs771148280 Chr.16: 46674905-46674905 on Build GRCh38
rs397701572 Chr.16: 46675011-46675012 on Build GRCh38
rs979434697 Chr.16: 46674851-46674851 on Build GRCh38
rs372867666 Chr.16: 46675092-46675093 on Build GRCh38
rs767511812 Chr.16: 46674781-46674783 on Build GRCh38
rs543178836 Chr.16: 46674781-46674781 on Build GRCh38
rs746705076 Chr.16: 46674662-46674663 on Build GRCh38
rs867030728 Chr.16: 46674661-46674661 on Build GRCh38
rs764047433 Chr.16: 46674663-46674663 on Build GRCh38
rs1030158643 Chr.16: 46674777-46674780 on Build GRCh38
rs566319063 Chr.16: 46674952-46674952 on Build GRCh38
rs77959480 Chr.16: 46675012-46675012 on Build GRCh38
rs747545277 Chr.16: 46674663-46674666 on Build GRCh38
rs770314763 Chr.16: 46674686-46674686 on Build GRCh38
rs569267771 Chr.16: 46674812-46674812 on Build GRCh38

View Full Product Details