The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001831.3 | NP_001822.3 | 3 - 4 | 436 | 65 |
NR_038335.1 | 3 - 4 | 571 | 65 | ||
NR_045494.1 | 3 - 4 | 430 | 65 | ||
XM_006716284.2 | XP_006716347.1 | 3 - 4 | 2342 | 65 | |
GenBank mRNA | AK093399.1 | - | 3 - 4 | 328 | 65 |
AK313870.1 | - | 3 - 4 | 325 | 65 | |
AY513288.1 | - | 3 - 4 | 315 | 65 | |
BC010514.1 | - | 3 - 4 | 318 | 65 | |
BC013221.1 | - | 2 - 3 | 251 | 65 | |
BC019588.2 | - | 2 - 3 | 286 | 65 | |
BC029972.1 | - | 3 - 4 | 318 | 65 | |
BG819541.1 | - | 3 - 4 | 525 | 65 | |
BP193735.1 | - | 3 - 4 | 430 | 65 | |
BX418836.2 | - | 3 - 4 | 555 | 65 | |
BX459023.2 | - | 3 - 4 | 309 | 65 | |
BX648414.1 | - | 2 - 3 | 1036 | 65 | |
DN990822.1 | - | 3 - 4 | 444 | 65 | |
EB385800.1 | - | 1 - 2 | 83 | 65 | |
J02908.1 | - | 1 - 2 | 168 | 65 | |
M25915.1 | - | 3 - 4 | 445 | 65 | |
M64722.1 | - | 3 - 4 | 302 | 65 | |
M74816.1 | - | 1 - 2 | 72 | 65 | |
X14723.1 | - | 2 - 3 | 297 | 65 |