Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71730481-71730976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1G1
Entrez Gene ID
164
Gene Name
adaptor related protein complex 1 gamma 1 subunit
Gene Aliases
ADTG, CLAPG1
UniGene
Hs.461253
Gene Chromosome Location
Chr.16: 71729000-71809073 on Build GRCh38
Transcripts
NM_001030007.1 NM_001128.5

SNP Detail

SNP ID Chromosome Location
rs3751817 Chr.16: 71730931-71730931 on Build GRCh38
rs749696225 Chr.16: 71730933-71730933 on Build GRCh38
rs113831755 Chr.16: 71730512-71730512 on Build GRCh38
rs747701192 Chr.16: 71730880-71730880 on Build GRCh38
rs753223551 Chr.16: 71730535-71730535 on Build GRCh38
rs959266812 Chr.16: 71730539-71730539 on Build GRCh38
rs909589814 Chr.16: 71730565-71730565 on Build GRCh38
rs959648929 Chr.16: 71730892-71730892 on Build GRCh38
rs780671496 Chr.16: 71730818-71730818 on Build GRCh38
rs116598525 Chr.16: 71730754-71730754 on Build GRCh38
rs746427720 Chr.16: 71730750-71730750 on Build GRCh38
rs1051424078 Chr.16: 71730651-71730651 on Build GRCh38
rs918355679 Chr.16: 71730953-71730954 on Build GRCh38
rs1003724273 Chr.16: 71730878-71730878 on Build GRCh38
rs990450698 Chr.16: 71730555-71730555 on Build GRCh38
rs145308652 Chr.16: 71730691-71730691 on Build GRCh38
rs933858569 Chr.16: 71730628-71730628 on Build GRCh38
rs540028962 Chr.16: 71730901-71730901 on Build GRCh38
rs376097642 Chr.16: 71730624-71730624 on Build GRCh38
rs542694062 Chr.16: 71730782-71730782 on Build GRCh38
rs1035643790 Chr.16: 71730891-71730891 on Build GRCh38
rs770758177 Chr.16: 71730951-71730951 on Build GRCh38
rs190958374 Chr.16: 71730829-71730829 on Build GRCh38
rs571226927 Chr.16: 71730607-71730607 on Build GRCh38
rs1021068947 Chr.16: 71730761-71730761 on Build GRCh38
rs918399270 Chr.16: 71730608-71730608 on Build GRCh38
rs941105275 Chr.16: 71730604-71730604 on Build GRCh38
rs150015997 Chr.16: 71730520-71730520 on Build GRCh38
rs184036267 Chr.16: 71730513-71730513 on Build GRCh38
rs546235794 Chr.16: 71730922-71730922 on Build GRCh38
rs573129642 Chr.16: 71730869-71730869 on Build GRCh38
rs769131565 Chr.16: 71730882-71730882 on Build GRCh38
rs866786192 Chr.16: 71730917-71730917 on Build GRCh38
rs149200383 Chr.16: 71730923-71730923 on Build GRCh38
rs74027296 Chr.16: 71730753-71730753 on Build GRCh38
rs1007933372 Chr.16: 71730709-71730709 on Build GRCh38

View Full Product Details