The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033553.2 | NP_291031.2 | 2 - 3 | 295 | 77 |
GenBank mRNA | M95174.1 | - | 2 - 3 | 287 | 77 |
M97496.1 | - | 2 - 3 | 291 | 77 |