Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HLA-DMB
- Entrez Gene ID
-
3109
- Gene Name
- major histocompatibility complex, class II, DM beta
- Gene Aliases
-
D6S221E, RING7
- Chromosome Location
-
Chr.6: 32934629 - 32941070 on Build GRCh38
- Gene Symbol
- HLA-DMA
- Entrez Gene ID
-
3108
- Gene Name
- major histocompatibility complex, class II, DM alpha
- Gene Aliases
-
D6S222E, DMA, HLADM, RING6
- Chromosome Location
-
Chr.6: 32948614 - 32953122 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AU100342.1 |
- |
2 - 3 |
241 |
100 |
View Full Product Details