Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 80607944-80608484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDYL2
Entrez Gene ID
124359
Gene Name
chromodomain protein, Y-like 2
Gene Aliases
PCCP1
UniGene
Hs.373908
Gene Chromosome Location
Chr.16: 80597899-80805043 on Build GRCh38
Transcripts
NM_152342.3 XM_011522866.1 XM_011522867.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063039 c.1287G>A p.S429S Chr.16: 80608167-80608167 on Build GRCh38
1609695 c.1305C>A p.T435T Chr.16: 80608149-80608149 on Build GRCh38
5754921 c.1360G>T p.V454L Chr.16: 80608094-80608094 on Build GRCh38
4262921 c.1355G>A p.S452N Chr.16: 80608099-80608099 on Build GRCh38
6625862 c.1345G>A p.A449T Chr.16: 80608109-80608109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777439337 Chr.16: 80608266-80608266 on Build GRCh38
rs771653331 Chr.16: 80608084-80608084 on Build GRCh38
rs753801497 Chr.16: 80608231-80608231 on Build GRCh38
rs750554842 Chr.16: 80608108-80608108 on Build GRCh38
rs758736300 Chr.16: 80608247-80608247 on Build GRCh38
rs775458776 Chr.16: 80608197-80608197 on Build GRCh38
rs534719734 Chr.16: 80608315-80608315 on Build GRCh38
rs565674343 Chr.16: 80608338-80608338 on Build GRCh38
rs772711195 Chr.16: 80608088-80608088 on Build GRCh38
rs930236067 Chr.16: 80608013-80608013 on Build GRCh38
rs989994238 Chr.16: 80608003-80608003 on Build GRCh38
rs777352705 Chr.16: 80608167-80608167 on Build GRCh38
rs1009437488 Chr.16: 80608388-80608388 on Build GRCh38
rs865804016 Chr.16: 80608136-80608136 on Build GRCh38
rs756721856 Chr.16: 80608055-80608055 on Build GRCh38
rs145676305 Chr.16: 80608094-80608094 on Build GRCh38
rs372996672 Chr.16: 80608119-80608119 on Build GRCh38
rs554516897 Chr.16: 80608363-80608363 on Build GRCh38
rs756240211 Chr.16: 80608113-80608113 on Build GRCh38
rs981123702 Chr.16: 80607985-80607985 on Build GRCh38
rs762788877 Chr.16: 80608199-80608199 on Build GRCh38
rs9940301 Chr.16: 80608009-80608009 on Build GRCh38
rs990497273 Chr.16: 80608201-80608201 on Build GRCh38
rs9933302 Chr.16: 80608034-80608034 on Build GRCh38
rs767900411 Chr.16: 80608200-80608200 on Build GRCh38
rs892292337 Chr.16: 80608350-80608350 on Build GRCh38
rs765690008 Chr.16: 80608238-80608238 on Build GRCh38
rs186378074 Chr.16: 80607999-80607999 on Build GRCh38
rs918623535 Chr.16: 80608359-80608359 on Build GRCh38
rs773537283 Chr.16: 80608221-80608221 on Build GRCh38
rs375996420 Chr.16: 80608161-80608161 on Build GRCh38
rs147583132 Chr.16: 80608140-80608140 on Build GRCh38
rs1044713868 Chr.16: 80608292-80608292 on Build GRCh38
rs536121602 Chr.16: 80608341-80608341 on Build GRCh38
rs770474965 Chr.16: 80608180-80608180 on Build GRCh38
rs776495063 Chr.16: 80608187-80608187 on Build GRCh38
rs113440630 Chr.16: 80608008-80608008 on Build GRCh38
rs148875027 Chr.16: 80608097-80608097 on Build GRCh38
rs181063533 Chr.16: 80608149-80608149 on Build GRCh38
rs543503956 Chr.16: 80608054-80608054 on Build GRCh38
rs541980100 Chr.16: 80608312-80608312 on Build GRCh38
rs931560667 Chr.16: 80608360-80608360 on Build GRCh38
rs542432411 Chr.16: 80608023-80608023 on Build GRCh38
rs748311359 Chr.16: 80608070-80608070 on Build GRCh38
rs745833710 Chr.16: 80608191-80608191 on Build GRCh38
rs369532733 Chr.16: 80608099-80608099 on Build GRCh38
rs953985177 Chr.16: 80608239-80608239 on Build GRCh38
rs75994894 Chr.16: 80608070-80608070 on Build GRCh38
rs142194846 Chr.16: 80608407-80608407 on Build GRCh38
rs767809619 Chr.16: 80608105-80608105 on Build GRCh38
rs185265872 Chr.16: 80608303-80608303 on Build GRCh38
rs948182516 Chr.16: 80608416-80608416 on Build GRCh38
rs114192586 Chr.16: 80608393-80608393 on Build GRCh38
rs191129809 Chr.16: 80608022-80608022 on Build GRCh38
rs1031555030 Chr.16: 80607992-80607992 on Build GRCh38
rs972699965 Chr.16: 80608327-80608327 on Build GRCh38
rs1021446086 Chr.16: 80608220-80608220 on Build GRCh38
rs896136445 Chr.16: 80608024-80608024 on Build GRCh38
rs904916344 Chr.16: 80608297-80608297 on Build GRCh38
rs890381627 Chr.16: 80608406-80608406 on Build GRCh38
rs912660934 Chr.16: 80608056-80608056 on Build GRCh38
rs532140476 Chr.16: 80608071-80608071 on Build GRCh38
rs556801871 Chr.16: 80608080-80608080 on Build GRCh38
rs753080970 Chr.16: 80608243-80608243 on Build GRCh38
rs780562299 Chr.16: 80608064-80608064 on Build GRCh38
rs904948514 Chr.16: 80608047-80608047 on Build GRCh38
rs942849359 Chr.16: 80607997-80607997 on Build GRCh38
rs760732086 Chr.16: 80608223-80608223 on Build GRCh38
rs765071393 Chr.16: 80608096-80608096 on Build GRCh38
rs116435329 Chr.16: 80608131-80608131 on Build GRCh38
rs577774106 Chr.16: 80608367-80608367 on Build GRCh38
rs918201160 Chr.16: 80608280-80608280 on Build GRCh38
rs949020346 Chr.16: 80608281-80608281 on Build GRCh38
rs377301254 Chr.16: 80608278-80608278 on Build GRCh38
rs146529911 Chr.16: 80608095-80608095 on Build GRCh38
rs117351184 Chr.16: 80608179-80608179 on Build GRCh38
rs748442310 Chr.16: 80608150-80608150 on Build GRCh38
rs1049148856 Chr.16: 80608376-80608376 on Build GRCh38
rs9940376 Chr.16: 80608039-80608039 on Build GRCh38
rs756806712 Chr.16: 80608284-80608284 on Build GRCh38
rs376314033 Chr.16: 80608079-80608079 on Build GRCh38
rs769555144 Chr.16: 80608195-80608195 on Build GRCh38
rs1030293611 Chr.16: 80608122-80608122 on Build GRCh38
rs1008210394 Chr.16: 80608444-80608445 on Build GRCh38
rs755512414 Chr.16: 80608235-80608235 on Build GRCh38
rs1046611326 Chr.16: 80608436-80608436 on Build GRCh38
rs750462998 Chr.16: 80608051-80608051 on Build GRCh38
rs74661853 Chr.16: 80608333-80608333 on Build GRCh38
rs766469320 Chr.16: 80608226-80608226 on Build GRCh38
rs865822442 Chr.16: 80608237-80608237 on Build GRCh38

View Full Product Details