Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 80803873-80804364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDYL2
Entrez Gene ID
124359
Gene Name
chromodomain protein, Y-like 2
Gene Aliases
PCCP1
UniGene
Hs.373908
Gene Chromosome Location
Chr.16: 80597899-80805043 on Build GRCh38
Transcripts
NM_152342.3 XM_011522866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5952889 c.24+4C>G p.? Chr.16: 80804146-80804146 on Build GRCh38
4645925 c.6T>G p.A2A Chr.16: 80804168-80804168 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs971312397 Chr.16: 80804038-80804038 on Build GRCh38
rs926860880 Chr.16: 80803920-80803920 on Build GRCh38
rs539952247 Chr.16: 80803995-80803995 on Build GRCh38
rs913553229 Chr.16: 80804090-80804099 on Build GRCh38
rs757491787 Chr.16: 80804140-80804140 on Build GRCh38
rs556049311 Chr.16: 80804219-80804219 on Build GRCh38
rs1011112693 Chr.16: 80804257-80804257 on Build GRCh38
rs1002800109 Chr.16: 80804254-80804254 on Build GRCh38
rs913669254 Chr.16: 80803927-80803927 on Build GRCh38
rs997625402 Chr.16: 80804255-80804268 on Build GRCh38
rs556158830 Chr.16: 80804301-80804301 on Build GRCh38
rs889556869 Chr.16: 80803969-80803969 on Build GRCh38
rs943332118 Chr.16: 80803970-80803970 on Build GRCh38
rs749809757 Chr.16: 80804175-80804175 on Build GRCh38
rs1039368626 Chr.16: 80803975-80803975 on Build GRCh38
rs534402864 Chr.16: 80804125-80804125 on Build GRCh38
rs183660830 Chr.16: 80803915-80803915 on Build GRCh38
rs1034367949 Chr.16: 80804048-80804048 on Build GRCh38
rs748504795 Chr.16: 80804206-80804206 on Build GRCh38
rs1051572065 Chr.16: 80803957-80803963 on Build GRCh38
rs1004298097 Chr.16: 80804019-80804019 on Build GRCh38
rs1004183370 Chr.16: 80804208-80804208 on Build GRCh38
rs981423495 Chr.16: 80803918-80803918 on Build GRCh38
rs543853720 Chr.16: 80804298-80804298 on Build GRCh38
rs936337907 Chr.16: 80803921-80803921 on Build GRCh38
rs911647785 Chr.16: 80804280-80804280 on Build GRCh38
rs987113006 Chr.16: 80804273-80804274 on Build GRCh38
rs980924332 Chr.16: 80804047-80804047 on Build GRCh38
rs887217351 Chr.16: 80804017-80804017 on Build GRCh38
rs866744496 Chr.16: 80804013-80804013 on Build GRCh38
rs750535092 Chr.16: 80804145-80804145 on Build GRCh38
rs1020610305 Chr.16: 80804259-80804260 on Build GRCh38
rs958256396 Chr.16: 80804054-80804055 on Build GRCh38
rs779942191 Chr.16: 80804158-80804158 on Build GRCh38
rs765019185 Chr.16: 80804128-80804128 on Build GRCh38
rs573442059 Chr.16: 80804108-80804108 on Build GRCh38
rs966355060 Chr.16: 80804272-80804272 on Build GRCh38
rs989137318 Chr.16: 80804061-80804061 on Build GRCh38
rs896850393 Chr.16: 80803990-80803990 on Build GRCh38
rs887099497 Chr.16: 80804195-80804195 on Build GRCh38
rs964464216 Chr.16: 80804286-80804286 on Build GRCh38
rs759332777 Chr.16: 80804121-80804121 on Build GRCh38
rs771887915 Chr.16: 80804222-80804222 on Build GRCh38
rs943645169 Chr.16: 80803973-80803973 on Build GRCh38
rs574322904 Chr.16: 80804228-80804228 on Build GRCh38
rs906594494 Chr.16: 80804247-80804247 on Build GRCh38
rs755503983 Chr.16: 80804184-80804184 on Build GRCh38
rs945117564 Chr.16: 80803951-80803951 on Build GRCh38
rs752449527 Chr.16: 80804129-80804129 on Build GRCh38
rs1041114742 Chr.16: 80803984-80803984 on Build GRCh38
rs1026592489 Chr.16: 80804004-80804004 on Build GRCh38
rs767696778 Chr.16: 80804144-80804144 on Build GRCh38
rs766292393 Chr.16: 80804099-80804099 on Build GRCh38
rs890282133 Chr.16: 80803957-80803970 on Build GRCh38
rs558307416 Chr.16: 80804104-80804104 on Build GRCh38
rs958306824 Chr.16: 80804255-80804255 on Build GRCh38
rs1025133963 Chr.16: 80803914-80803914 on Build GRCh38
rs901986824 Chr.16: 80804133-80804134 on Build GRCh38
rs1025175800 Chr.16: 80804025-80804025 on Build GRCh38
rs779446449 Chr.16: 80804199-80804199 on Build GRCh38
rs989085162 Chr.16: 80803926-80803926 on Build GRCh38
rs995524605 Chr.16: 80804012-80804020 on Build GRCh38
rs930533033 Chr.16: 80804179-80804179 on Build GRCh38
rs899041883 Chr.16: 80804011-80804012 on Build GRCh38
rs753798894 Chr.16: 80804111-80804111 on Build GRCh38

View Full Product Details