The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002318.2 | NP_002309.1 | 10 - 11 | 2222 | 62 |
GenBank mRNA | AF117949.1 | - | 7 - 8 | 1352 | 62 |
AK222477.1 | - | 10 - 11 | 2133 | 62 | |
AK293292.1 | - | 7 - 8 | 1048 | 62 | |
AK312266.1 | - | 10 - 11 | 2134 | 62 | |
BC000594.2 | - | 10 - 11 | 2092 | 62 | |
KF928961.1 | - | 9 - 10 | 1883 | 62 | |
KF975332.1 | - | 9 - 10 | 1823 | 62 | |
U89942.1 | - | 10 - 11 | 2130 | 62 |