The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270987.1 | NP_001257916.1 | 8 - 9 | 930 | 73 |
NM_002342.2 | NP_002333.1 | 8 - 9 | 1125 | 73 | |
GenBank mRNA | AK027080.1 | - | 8 - 9 | 990 | 73 |
AK095418.1 | - | 9 - 10 | 954 | 73 | |
AK293187.1 | - | 8 - 9 | 869 | 73 | |
BC026262.1 | - | 8 - 9 | 997 | 73 | |
L04270.1 | - | 8 - 9 | 967 | 73 |