Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOG
Entrez Gene ID
4340
Gene Name
myelin oligodendrocyte glycoprotein
Gene Aliases
BTN6, BTNL11, MOGIG2, NRCLP7
Chromosome Location
Chr.6: 29656981 - 29672372 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002433.4 NP_002424.3 5 - 6 817 72
NM_206809.3 NP_996532.2 5 - 6 817 72
NM_206814.5 NP_996537.3 4 - 5 469 72
XM_005249131.3 XP_005249188.1 5 - 6 806 72
GenBank mRNA AB209815.1 - 5 - 6 727 72
AK295213.1 - 6 - 7 638 72
AK308949.1 - 4 - 5 447 72
AK312892.1 - 5 - 6 703 72
AY566852.1 - 5 - 6 588 72
BC035938.1 - 5 - 6 706 72
U18798.1 - 5 - 6 588 72
U18800.2 - 4 - 5 240 72
U18801.1 - 5 - 6 588 72
U64564.1 - 5 - 6 750 72
U64565.1 - 4 - 5 501 72
U64570.1 - 5 - 6 639 72
U64571.1 - 5 - 6 639 72
X74511.1 - 5 - 6 588 72

View Full Product Details