Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NMBR
Entrez Gene ID
4829
Gene Name
neuromedin B receptor
Gene Aliases
BB1, BB1R, NMB-R
Chromosome Location
Chr.6: 142074484 - 142147205 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002511.3 NP_002502.2 1 - 2 568 82
GenBank mRNA BC095542.1 - 1 - 2 428 82
GU727635.1 - 1 - 2 427 82
M73482.1 - 1 - 2 566 82

View Full Product Details