Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1005773-1006268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABR
Entrez Gene ID
29
Gene Name
active BCR-related
Gene Aliases
MDB
UniGene
Hs.159306
Gene Chromosome Location
Chr.17: 1003518-1229713 on Build GRCh38
Transcripts
NM_001092.4 NM_001159746.2 NM_001256847.2 NM_001282149.1 NM_001322840.1 NM_001322841.1 NM_021962.4 XM_011523810.2 XM_011523812.2 XM_011523813.2 XM_011523814.2 XM_011523815.2 XM_017024539.1 XM_017024540.1 XM_017024542.1 XM_017024543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4929100 c.2572G>A p.D858N Chr.17: 1006088-1006088 on Build GRCh38
4612763 c.2415delC p.I806fs*>17 Chr.17: 1006134-1006134 on Build GRCh38
4929101 c.2461G>A p.D821N Chr.17: 1006088-1006088 on Build GRCh38
1387675 c.2526delC p.I843fs*>17 Chr.17: 1006134-1006134 on Build GRCh38
112051 c.2526_2527insC p.I843fs*>18 Chr.17: 1006133-1006134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757968067 Chr.17: 1006188-1006188 on Build GRCh38
rs762668237 Chr.17: 1006173-1006173 on Build GRCh38
rs545347241 Chr.17: 1006084-1006084 on Build GRCh38
rs756316337 Chr.17: 1006051-1006051 on Build GRCh38
rs191116738 Chr.17: 1006140-1006140 on Build GRCh38
rs955841245 Chr.17: 1005926-1005926 on Build GRCh38
rs752452866 Chr.17: 1006046-1006046 on Build GRCh38
rs370073889 Chr.17: 1006095-1006095 on Build GRCh38
rs865958326 Chr.17: 1006085-1006085 on Build GRCh38
rs751542338 Chr.17: 1006025-1006025 on Build GRCh38
rs35089910 Chr.17: 1006086-1006086 on Build GRCh38
rs772988592 Chr.17: 1006079-1006079 on Build GRCh38
rs920684863 Chr.17: 1006033-1006033 on Build GRCh38
rs755872294 Chr.17: 1006108-1006108 on Build GRCh38
rs769136114 Chr.17: 1006135-1006135 on Build GRCh38
rs936756832 Chr.17: 1005936-1005936 on Build GRCh38
rs773962381 Chr.17: 1006189-1006189 on Build GRCh38
rs368718355 Chr.17: 1006218-1006218 on Build GRCh38
rs766047001 Chr.17: 1006000-1006000 on Build GRCh38
rs772478821 Chr.17: 1006068-1006068 on Build GRCh38
rs912801859 Chr.17: 1005906-1005906 on Build GRCh38
rs1004719955 Chr.17: 1006042-1006042 on Build GRCh38
rs542031344 Chr.17: 1006026-1006026 on Build GRCh38
rs750897785 Chr.17: 1006133-1006134 on Build GRCh38
rs978314955 Chr.17: 1006125-1006125 on Build GRCh38
rs80119159 Chr.17: 1006113-1006113 on Build GRCh38
rs749888317 Chr.17: 1005967-1005967 on Build GRCh38
rs373385043 Chr.17: 1006112-1006112 on Build GRCh38
rs199810449 Chr.17: 1006057-1006057 on Build GRCh38
rs745965118 Chr.17: 1006128-1006128 on Build GRCh38
rs973478351 Chr.17: 1005992-1005993 on Build GRCh38
rs983074294 Chr.17: 1005833-1005833 on Build GRCh38
rs765448631 Chr.17: 1006220-1006220 on Build GRCh38
rs1025294170 Chr.17: 1005819-1005819 on Build GRCh38
rs895004380 Chr.17: 1005999-1005999 on Build GRCh38
rs965620977 Chr.17: 1006111-1006111 on Build GRCh38
rs140781589 Chr.17: 1005796-1005796 on Build GRCh38
rs775885012 Chr.17: 1006076-1006076 on Build GRCh38
rs752145953 Chr.17: 1006092-1006094 on Build GRCh38
rs758976634 Chr.17: 1006190-1006190 on Build GRCh38
rs73978318 Chr.17: 1005916-1005916 on Build GRCh38
rs752467321 Chr.17: 1006104-1006104 on Build GRCh38
rs566969388 Chr.17: 1005824-1005824 on Build GRCh38
rs772563422 Chr.17: 1006129-1006129 on Build GRCh38
rs754276012 Chr.17: 1005919-1005919 on Build GRCh38
rs185224914 Chr.17: 1005803-1005803 on Build GRCh38
rs371379523 Chr.17: 1006056-1006056 on Build GRCh38
rs775278570 Chr.17: 1006209-1006209 on Build GRCh38
rs574573769 Chr.17: 1005933-1005933 on Build GRCh38
rs779030703 Chr.17: 1006061-1006061 on Build GRCh38
rs963102589 Chr.17: 1005850-1005850 on Build GRCh38
rs1046664831 Chr.17: 1006225-1006225 on Build GRCh38
rs200944927 Chr.17: 1005880-1005880 on Build GRCh38
rs537199930 Chr.17: 1005884-1005885 on Build GRCh38
rs62068292 Chr.17: 1006167-1006167 on Build GRCh38
rs762142683 Chr.17: 1006057-1006058 on Build GRCh38
rs372014888 Chr.17: 1006217-1006217 on Build GRCh38
rs900171954 Chr.17: 1006062-1006062 on Build GRCh38
rs920758527 Chr.17: 1005832-1005832 on Build GRCh38
rs926593577 Chr.17: 1005990-1005990 on Build GRCh38
rs559958422 Chr.17: 1006027-1006027 on Build GRCh38
rs1046344684 Chr.17: 1006021-1006021 on Build GRCh38
rs544558642 Chr.17: 1005913-1005913 on Build GRCh38
rs1008538170 Chr.17: 1005858-1005858 on Build GRCh38
rs757997639 Chr.17: 1005982-1005982 on Build GRCh38
rs985752125 Chr.17: 1005883-1005883 on Build GRCh38
rs1054081 Chr.17: 1005896-1005896 on Build GRCh38
rs375629677 Chr.17: 1006089-1006089 on Build GRCh38
rs780437158 Chr.17: 1006134-1006134 on Build GRCh38
rs533898378 Chr.17: 1005835-1005835 on Build GRCh38
rs562178600 Chr.17: 1005855-1005855 on Build GRCh38
rs1053859996 Chr.17: 1005976-1005976 on Build GRCh38
rs765717611 Chr.17: 1006130-1006130 on Build GRCh38
rs1042421160 Chr.17: 1005932-1005932 on Build GRCh38
rs758462978 Chr.17: 1006134-1006134 on Build GRCh38
rs767434137 Chr.17: 1006203-1006203 on Build GRCh38
rs906344119 Chr.17: 1006235-1006235 on Build GRCh38
rs770983190 Chr.17: 1006060-1006060 on Build GRCh38
rs757511977 Chr.17: 1006124-1006124 on Build GRCh38
rs2586306 Chr.17: 1006211-1006211 on Build GRCh38
rs868321988 Chr.17: 1006107-1006107 on Build GRCh38
rs929452368 Chr.17: 1006041-1006041 on Build GRCh38
rs867637236 Chr.17: 1006138-1006138 on Build GRCh38
rs374118537 Chr.17: 1006179-1006179 on Build GRCh38
rs751168415 Chr.17: 1006088-1006088 on Build GRCh38
rs920104574 Chr.17: 1005869-1005869 on Build GRCh38
rs796321696 Chr.17: 1005959-1005959 on Build GRCh38
rs1012491322 Chr.17: 1006004-1006004 on Build GRCh38
rs760952801 Chr.17: 1006077-1006077 on Build GRCh38

View Full Product Details