Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1012843-1013332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABR
Entrez Gene ID
29
Gene Name
active BCR-related
Gene Aliases
MDB
UniGene
Hs.159306
Gene Chromosome Location
Chr.17: 1003518-1229713 on Build GRCh38
Transcripts
NM_001092.4 NM_001159746.2 NM_001256847.2 NM_001282149.1 NM_001322840.1 NM_001322841.1 NM_021962.4 XM_011523810.2 XM_011523812.2 XM_011523813.2 XM_011523814.2 XM_011523815.2 XM_017024539.1 XM_017024540.1 XM_017024542.1 XM_017024543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1646867 c.1722C>G p.D574E Chr.17: 1013123-1013123 on Build GRCh38
5387777 c.1720G>A p.D574N Chr.17: 1013125-1013125 on Build GRCh38
708487 c.1833C>G p.D611E Chr.17: 1013123-1013123 on Build GRCh38
6591306 c.1737C>T p.N579N Chr.17: 1013108-1013108 on Build GRCh38
4748376 c.1834G>A p.V612M Chr.17: 1013122-1013122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906010066 Chr.17: 1013141-1013141 on Build GRCh38
rs962481847 Chr.17: 1013289-1013289 on Build GRCh38
rs369321976 Chr.17: 1013193-1013193 on Build GRCh38
rs777952601 Chr.17: 1013094-1013094 on Build GRCh38
rs376515484 Chr.17: 1013068-1013068 on Build GRCh38
rs769517467 Chr.17: 1013202-1013202 on Build GRCh38
rs958780050 Chr.17: 1013256-1013256 on Build GRCh38
rs567410742 Chr.17: 1013009-1013009 on Build GRCh38
rs571299658 Chr.17: 1012880-1012880 on Build GRCh38
rs184555399 Chr.17: 1013269-1013269 on Build GRCh38
rs1016519572 Chr.17: 1012930-1012930 on Build GRCh38
rs769285645 Chr.17: 1013136-1013136 on Build GRCh38
rs759310881 Chr.17: 1013149-1013149 on Build GRCh38
rs979241138 Chr.17: 1013274-1013274 on Build GRCh38
rs970641525 Chr.17: 1012999-1012999 on Build GRCh38
rs955223733 Chr.17: 1013240-1013240 on Build GRCh38
rs534908325 Chr.17: 1013227-1013227 on Build GRCh38
rs984301302 Chr.17: 1013000-1013000 on Build GRCh38
rs937235457 Chr.17: 1013006-1013006 on Build GRCh38
rs773956624 Chr.17: 1013148-1013148 on Build GRCh38
rs771048550 Chr.17: 1012874-1012874 on Build GRCh38
rs138764017 Chr.17: 1013056-1013056 on Build GRCh38
rs954831277 Chr.17: 1012954-1012954 on Build GRCh38
rs200217863 Chr.17: 1013097-1013097 on Build GRCh38
rs370908585 Chr.17: 1013306-1013306 on Build GRCh38
rs879165914 Chr.17: 1013244-1013244 on Build GRCh38
rs141106809 Chr.17: 1013108-1013108 on Build GRCh38
rs116071101 Chr.17: 1013180-1013180 on Build GRCh38
rs538615360 Chr.17: 1013270-1013270 on Build GRCh38
rs575444428 Chr.17: 1013284-1013284 on Build GRCh38
rs200395437 Chr.17: 1013127-1013127 on Build GRCh38
rs557365940 Chr.17: 1013283-1013283 on Build GRCh38
rs776417075 Chr.17: 1013109-1013109 on Build GRCh38
rs386794316 Chr.17: 1013205-1013206 on Build GRCh38
rs373518659 Chr.17: 1013057-1013057 on Build GRCh38
rs929763474 Chr.17: 1013032-1013032 on Build GRCh38
rs779489922 Chr.17: 1013184-1013184 on Build GRCh38
rs535037999 Chr.17: 1013084-1013084 on Build GRCh38
rs192182780 Chr.17: 1013257-1013257 on Build GRCh38
rs927214651 Chr.17: 1013001-1013001 on Build GRCh38
rs747841898 Chr.17: 1013122-1013122 on Build GRCh38
rs1052446031 Chr.17: 1013219-1013219 on Build GRCh38
rs770902392 Chr.17: 1013139-1013139 on Build GRCh38
rs1037886984 Chr.17: 1013192-1013192 on Build GRCh38
rs989290028 Chr.17: 1012963-1012963 on Build GRCh38
rs4968074 Chr.17: 1013226-1013226 on Build GRCh38
rs760937665 Chr.17: 1013161-1013161 on Build GRCh38
rs1014794598 Chr.17: 1012994-1012994 on Build GRCh38
rs780610506 Chr.17: 1013199-1013199 on Build GRCh38
rs751035631 Chr.17: 1013190-1013190 on Build GRCh38
rs753985892 Chr.17: 1013174-1013174 on Build GRCh38
rs764035976 Chr.17: 1013170-1013170 on Build GRCh38
rs985187913 Chr.17: 1013257-1013260 on Build GRCh38
rs150193335 Chr.17: 1013031-1013031 on Build GRCh38
rs926994472 Chr.17: 1012871-1012871 on Build GRCh38
rs922164597 Chr.17: 1013303-1013303 on Build GRCh38
rs776215842 Chr.17: 1013055-1013055 on Build GRCh38
rs746412503 Chr.17: 1013296-1013296 on Build GRCh38
rs994292426 Chr.17: 1012884-1012884 on Build GRCh38
rs9894835 Chr.17: 1013206-1013206 on Build GRCh38
rs886642543 Chr.17: 1013035-1013035 on Build GRCh38
rs979719496 Chr.17: 1012945-1012955 on Build GRCh38
rs138761504 Chr.17: 1013150-1013150 on Build GRCh38
rs1000221374 Chr.17: 1013245-1013245 on Build GRCh38
rs528243183 Chr.17: 1012866-1012866 on Build GRCh38
rs367556057 Chr.17: 1012895-1012895 on Build GRCh38
rs201108964 Chr.17: 1013203-1013203 on Build GRCh38
rs147397030 Chr.17: 1013123-1013123 on Build GRCh38
rs1006519074 Chr.17: 1012910-1012910 on Build GRCh38
rs370499754 Chr.17: 1013069-1013069 on Build GRCh38
rs968513362 Chr.17: 1012987-1012987 on Build GRCh38
rs374210585 Chr.17: 1013079-1013079 on Build GRCh38
rs752886863 Chr.17: 1013159-1013159 on Build GRCh38
rs1020217842 Chr.17: 1013230-1013230 on Build GRCh38
rs922366815 Chr.17: 1012879-1012879 on Build GRCh38
rs577232327 Chr.17: 1012996-1012996 on Build GRCh38
rs368841815 Chr.17: 1013131-1013131 on Build GRCh38
rs373944590 Chr.17: 1013204-1013204 on Build GRCh38
rs111972847 Chr.17: 1012935-1012935 on Build GRCh38
rs913091518 Chr.17: 1012986-1012986 on Build GRCh38
rs886586961 Chr.17: 1012897-1012897 on Build GRCh38
rs73283430 Chr.17: 1013060-1013060 on Build GRCh38
rs746240169 Chr.17: 1013107-1013107 on Build GRCh38
rs747749929 Chr.17: 1013200-1013200 on Build GRCh38
rs537198980 Chr.17: 1012974-1012974 on Build GRCh38
rs759511129 Chr.17: 1013212-1013212 on Build GRCh38
rs2241924 Chr.17: 1012946-1012946 on Build GRCh38
rs910975949 Chr.17: 1013259-1013259 on Build GRCh38
rs751682209 Chr.17: 1013186-1013189 on Build GRCh38
rs549148622 Chr.17: 1013223-1013223 on Build GRCh38
rs78616171 Chr.17: 1013285-1013285 on Build GRCh38
rs200093564 Chr.17: 1013076-1013076 on Build GRCh38
rs546611412 Chr.17: 1012869-1012869 on Build GRCh38
rs937014699 Chr.17: 1012873-1012873 on Build GRCh38
rs909522035 Chr.17: 1013286-1013286 on Build GRCh38
rs755123591 Chr.17: 1013075-1013075 on Build GRCh38
rs1034167240 Chr.17: 1013247-1013247 on Build GRCh38
rs904628466 Chr.17: 1013238-1013238 on Build GRCh38
rs1036715082 Chr.17: 1012872-1012872 on Build GRCh38
rs187475230 Chr.17: 1013033-1013033 on Build GRCh38
rs184277768 Chr.17: 1012947-1012947 on Build GRCh38
rs762996568 Chr.17: 1013064-1013064 on Build GRCh38
rs774269769 Chr.17: 1013209-1013209 on Build GRCh38
rs767030779 Chr.17: 1013103-1013103 on Build GRCh38
rs916885342 Chr.17: 1013013-1013013 on Build GRCh38
rs143125780 Chr.17: 1013098-1013098 on Build GRCh38
rs1031706999 Chr.17: 1012977-1012977 on Build GRCh38

View Full Product Details