Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNMT
Entrez Gene ID
5409
Gene Name
phenylethanolamine N-methyltransferase
Gene Aliases
PENT, PNMTase
Chromosome Location
Chr.17: 39667981 - 39670475 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002686.4 NP_002677.1 1 - 2 228 86
GenBank mRNA BC037246.1 - 1 - 2 427 86
CR541815.1 - 1 - 2 205 86
J03727.1 - 1 - 2 211 86

View Full Product Details