The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002899.3 | NP_002890.2 | 2 - 3 | 547 | 81 |
GenBank mRNA | AK290315.1 | - | 2 - 3 | 354 | 81 |
BC121052.2 | - | 2 - 3 | 305 | 81 | |
BU734286.1 | - | 2 - 3 | 81 | ||
CR541979.1 | - | 2 - 3 | 250 | 81 | |
CR542005.1 | - | 2 - 3 | 250 | 81 | |
M11433.1 | - | 2 - 3 | 375 | 81 | |
M36809.1 | - | 2 - 3 | 322 | 81 |