The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002926.3 | NP_002917.1 | 4 - 5 | 2926 | 75 |
NM_198229.2 | NP_937872.1 | 4 - 5 | 2926 | 75 | |
XM_006713905.1 | XP_006713968.1 | 4 - 5 | 2245 | 75 | |
XM_006713906.2 | XP_006713969.1 | 4 - 5 | 2245 | 75 | |
XM_011513543.2 | XP_011511845.1 | 3 - 4 | 201 | 75 | |
XM_017008529.1 | XP_016864018.1 | 4 - 5 | 3495 | 75 | |
GenBank mRNA | AF030111.1 | - | 4 - 5 | 2925 | 75 |
AF035152.1 | - | 3 - 4 | 2076 | 75 | |
AF464737.1 | - | 3 - 4 | 301 | 75 | |
AK310999.1 | - | 3 - 4 | 213 | 75 | |
BC028154.1 | - | 4 - 5 | 2188 | 75 | |
BC118594.1 | - | 3 - 4 | 292 | 75 |