The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002931.3 | NP_002922.2 | 6 - 7 | 1316 | 112 |
GenBank mRNA | AK289355.1 | - | 6 - 7 | 1325 | 112 |
AK301378.1 | - | 6 - 7 | 1259 | 112 | |
BC002922.2 | - | 6 - 7 | 1311 | 112 | |
BC051866.2 | - | 6 - 7 | 1309 | 112 | |
BT007352.1 | - | 4 - 5 | 1035 | 112 | |
Z14000.1 | - | 4 - 5 | 1110 | 112 |