The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002976.3 | NP_002967.2 | 10 - 11 | 1421 | 109 |
NR_045628.1 | 10 - 11 | 1421 | 109 | ||
XM_006712680.2 | XP_006712743.1 | 10 - 11 | 1490 | 109 | |
XM_006712681.3 | XP_006712744.1 | 11 - 12 | 1715 | 109 | |
XM_006712682.3 | XP_006712745.1 | 12 - 13 | 1840 | 109 | |
XM_011511615.2 | XP_011509917.1 | 12 - 13 | 1786 | 109 | |
XM_017004667.1 | XP_016860156.1 | 12 - 13 | 1759 | 109 | |
GenBank mRNA | BC141864.1 | - | 10 - 11 | 1423 | 109 |
M91556.1 | - | 10 - 11 | 1430 | 109 |