Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7217037-7217532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADVL
Entrez Gene ID
37
Gene Name
acyl-CoA dehydrogenase, very long chain
Gene Aliases
ACAD6, LCACD, VLCAD
UniGene
Hs.437178
Gene Chromosome Location
Chr.17: 7217125-7225267 on Build GRCh38
Transcripts
NM_001270447.1 NM_001128827.2 NM_001321074.1 NM_001321075.1 NM_001365.4 NR_135527.1 XM_005256491.1 XM_011523698.1 XM_011523699.2 XM_017024288.1 XM_017024289.1

Gene Symbol
DLG4
Entrez Gene ID
1742
Gene Name
discs large MAGUK scaffold protein 4
Gene Aliases
PSD95, SAP-90, SAP90
UniGene
Hs.463928
Gene Chromosome Location
Chr.17: 7189890-7220050 on Build GRCh38
Transcripts
NM_001270447.1 NM_001128827.2 NM_001321074.1 NM_001321075.1 NM_001365.4 NR_135527.1 XM_005256491.1 XM_011523698.1 XM_011523699.2 XM_017024288.1 XM_017024289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
983809 c.12_13delCT p.C5fs*12 Chr.17: 7217135-7217136 on Build GRCh38
1385919 c.3G>A p.M1I Chr.17: 7217145-7217145 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs965064636 Chr.17: 7217471-7217471 on Build GRCh38
rs1044810173 Chr.17: 7217153-7217153 on Build GRCh38
rs753669668 Chr.17: 7217179-7217179 on Build GRCh38
rs923946168 Chr.17: 7217464-7217464 on Build GRCh38
rs1026312176 Chr.17: 7217409-7217409 on Build GRCh38
rs755662999 Chr.17: 7217099-7217100 on Build GRCh38
rs1022488680 Chr.17: 7217393-7217393 on Build GRCh38
rs961029201 Chr.17: 7217469-7217469 on Build GRCh38
rs978610306 Chr.17: 7217419-7217419 on Build GRCh38
rs532873127 Chr.17: 7217241-7217241 on Build GRCh38
rs549454733 Chr.17: 7217327-7217327 on Build GRCh38
rs994279885 Chr.17: 7217209-7217209 on Build GRCh38
rs899233625 Chr.17: 7217216-7217216 on Build GRCh38
rs897369799 Chr.17: 7217195-7217195 on Build GRCh38
rs998446241 Chr.17: 7217248-7217248 on Build GRCh38
rs1029953321 Chr.17: 7217282-7217282 on Build GRCh38
rs968129768 Chr.17: 7217413-7217413 on Build GRCh38
rs1018364474 Chr.17: 7217378-7217378 on Build GRCh38
rs888212677 Chr.17: 7217259-7217259 on Build GRCh38
rs903474904 Chr.17: 7217101-7217101 on Build GRCh38
rs753057103 Chr.17: 7217064-7217064 on Build GRCh38
rs992694235 Chr.17: 7217475-7217475 on Build GRCh38
rs760680786 Chr.17: 7217106-7217106 on Build GRCh38
rs938941588 Chr.17: 7217181-7217181 on Build GRCh38
rs934919111 Chr.17: 7217105-7217105 on Build GRCh38
rs1006553926 Chr.17: 7217306-7217310 on Build GRCh38
rs923802740 Chr.17: 7217097-7217097 on Build GRCh38
rs569357655 Chr.17: 7217337-7217337 on Build GRCh38
rs995270272 Chr.17: 7217250-7217250 on Build GRCh38
rs757028100 Chr.17: 7217206-7217206 on Build GRCh38
rs946405930 Chr.17: 7217071-7217071 on Build GRCh38
rs954286284 Chr.17: 7217367-7217367 on Build GRCh38
rs2074220 Chr.17: 7217100-7217100 on Build GRCh38
rs1007024757 Chr.17: 7217387-7217387 on Build GRCh38
rs1007706923 Chr.17: 7217163-7217163 on Build GRCh38
rs1017714299 Chr.17: 7217186-7217186 on Build GRCh38
rs914647414 Chr.17: 7217104-7217104 on Build GRCh38
rs777384926 Chr.17: 7217100-7217100 on Build GRCh38
rs763975299 Chr.17: 7217159-7217159 on Build GRCh38
rs766483452 Chr.17: 7217077-7217077 on Build GRCh38
rs1051981436 Chr.17: 7217121-7217121 on Build GRCh38

View Full Product Details