The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128174.2 | NP_001121646.1 | 4 - 5 | 1617 | 150 |
NM_001322112.1 | NP_001309041.1 | 5 - 6 | 1720 | 150 | |
NM_001322113.1 | NP_001309042.1 | 4 - 5 | 1700 | 150 | |
NM_001322114.1 | NP_001309043.1 | 4 - 5 | 1546 | 150 | |
NM_003360.4 | NP_003351.2 | 3 - 4 | 1556 | 150 | |
GenBank mRNA | AK127970.1 | - | 4 - 5 | 1547 | 150 |
AK225763.1 | - | 4 - 5 | 1210 | 150 | |
AK294438.1 | - | 3 - 4 | 976 | 150 | |
BC075069.2 | - | 4 - 5 | 1089 | 150 | |
U30930.1 | - | 3 - 4 | 1557 | 150 | |
U62899.1 | - | 4 - 5 | 1268 | 150 |