Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30803489-30804032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLF3
Entrez Gene ID
51379
Gene Name
cytokine receptor like factor 3
Gene Aliases
CREME-9, CREME9, CRLM9, CYTOR4, FRWS, p48.2
UniGene
Hs.649372
Gene Chromosome Location
Chr.17: 30782684-30824760 on Build GRCh38
Transcripts
NM_015986.3 NR_073118.1 XM_011524879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3179332 c.303C>T p.H101H Chr.17: 30803935-30803935 on Build GRCh38
1630119 c.225G>A p.V75V Chr.17: 30804013-30804013 on Build GRCh38
7194830 c.220T>C p.L74L Chr.17: 30804018-30804018 on Build GRCh38
977331 c.217C>T p.R73* Chr.17: 30804021-30804021 on Build GRCh38
6282049 c.321G>A p.E107E Chr.17: 30803917-30803917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891094304 Chr.17: 30803636-30803636 on Build GRCh38
rs573779590 Chr.17: 30803620-30803620 on Build GRCh38
rs776013029 Chr.17: 30803852-30803852 on Build GRCh38
rs758593649 Chr.17: 30803914-30803914 on Build GRCh38
rs558124617 Chr.17: 30803582-30803582 on Build GRCh38
rs936883465 Chr.17: 30803544-30803544 on Build GRCh38
rs770510980 Chr.17: 30803964-30803964 on Build GRCh38
rs573593081 Chr.17: 30803575-30803575 on Build GRCh38
rs1029667830 Chr.17: 30803629-30803629 on Build GRCh38
rs886263453 Chr.17: 30803894-30803894 on Build GRCh38
rs745887448 Chr.17: 30803917-30803917 on Build GRCh38
rs1054085364 Chr.17: 30803551-30803551 on Build GRCh38
rs756747028 Chr.17: 30803927-30803927 on Build GRCh38
rs563409583 Chr.17: 30803760-30803760 on Build GRCh38
rs769084122 Chr.17: 30803951-30803951 on Build GRCh38
rs545305277 Chr.17: 30803727-30803727 on Build GRCh38
rs202141263 Chr.17: 30803897-30803897 on Build GRCh38
rs542630612 Chr.17: 30803824-30803824 on Build GRCh38
rs185374807 Chr.17: 30803689-30803689 on Build GRCh38
rs1002638135 Chr.17: 30803590-30803590 on Build GRCh38
rs112220518 Chr.17: 30803976-30803976 on Build GRCh38
rs778573807 Chr.17: 30803527-30803527 on Build GRCh38
rs892787478 Chr.17: 30803553-30803553 on Build GRCh38
rs577716055 Chr.17: 30803712-30803712 on Build GRCh38
rs1030306173 Chr.17: 30803696-30803696 on Build GRCh38
rs375223178 Chr.17: 30803875-30803875 on Build GRCh38
rs749659965 Chr.17: 30803935-30803935 on Build GRCh38
rs760666339 Chr.17: 30803664-30803664 on Build GRCh38
rs561008148 Chr.17: 30803826-30803826 on Build GRCh38
rs990371268 Chr.17: 30803697-30803697 on Build GRCh38
rs886277203 Chr.17: 30803607-30803607 on Build GRCh38
rs761148577 Chr.17: 30803783-30803783 on Build GRCh38
rs748192336 Chr.17: 30803886-30803887 on Build GRCh38
rs752553933 Chr.17: 30803884-30803884 on Build GRCh38
rs1006988623 Chr.17: 30803667-30803667 on Build GRCh38
rs3794796 Chr.17: 30803862-30803862 on Build GRCh38
rs780706405 Chr.17: 30803928-30803928 on Build GRCh38
rs570045635 Chr.17: 30803594-30803594 on Build GRCh38
rs868532519 Chr.17: 30803806-30803806 on Build GRCh38
rs765170936 Chr.17: 30803698-30803698 on Build GRCh38
rs924069839 Chr.17: 30803771-30803771 on Build GRCh38
rs373445802 Chr.17: 30803898-30803898 on Build GRCh38
rs758180590 Chr.17: 30803892-30803892 on Build GRCh38
rs770361080 Chr.17: 30803851-30803851 on Build GRCh38
rs144562177 Chr.17: 30803979-30803979 on Build GRCh38
rs978153728 Chr.17: 30803770-30803770 on Build GRCh38
rs142019110 Chr.17: 30803762-30803762 on Build GRCh38
rs981111544 Chr.17: 30803725-30803725 on Build GRCh38
rs201388613 Chr.17: 30803913-30803913 on Build GRCh38
rs1026118535 Chr.17: 30803598-30803598 on Build GRCh38
rs1006549333 Chr.17: 30803657-30803657 on Build GRCh38
rs189636682 Chr.17: 30803906-30803906 on Build GRCh38
rs375192623 Chr.17: 30803878-30803878 on Build GRCh38
rs924123624 Chr.17: 30803521-30803521 on Build GRCh38
rs113370586 Chr.17: 30803864-30803864 on Build GRCh38
rs765941251 Chr.17: 30803870-30803870 on Build GRCh38
rs957246498 Chr.17: 30803694-30803694 on Build GRCh38
rs201213537 Chr.17: 30803855-30803855 on Build GRCh38
rs996061068 Chr.17: 30804005-30804005 on Build GRCh38
rs774536161 Chr.17: 30803956-30803956 on Build GRCh38
rs996386276 Chr.17: 30803749-30803749 on Build GRCh38
rs1016215754 Chr.17: 30803662-30803662 on Build GRCh38
rs568894417 Chr.17: 30803536-30803537 on Build GRCh38
rs760193135 Chr.17: 30803865-30803865 on Build GRCh38
rs1020228851 Chr.17: 30803706-30803706 on Build GRCh38
rs775284587 Chr.17: 30803588-30803588 on Build GRCh38
rs765144043 Chr.17: 30803879-30803879 on Build GRCh38
rs528194920 Chr.17: 30803837-30803837 on Build GRCh38
rs540867083 Chr.17: 30803671-30803671 on Build GRCh38
rs967690862 Chr.17: 30803714-30803714 on Build GRCh38
rs555467106 Chr.17: 30803617-30803617 on Build GRCh38
rs537016199 Chr.17: 30803595-30803595 on Build GRCh38
rs766686611 Chr.17: 30803944-30803944 on Build GRCh38

View Full Product Details