The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000046.3 | NP_000037.2 | 6 - 7 | 2503 | 57 |
XM_011543390.1 | XP_011541692.1 | 7 - 8 | 1716 | 57 | |
GenBank mRNA | AK314903.1 | - | 6 - 7 | 1288 | 57 |
M32373.1 | - | 6 - 7 | 1776 | 57 |