The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017006967.1 | XP_016862456.1 | 2 - 3 | 1678 | 64 |
GenBank mRNA | AK292063.1 | - | 2 - 3 | 1636 | 64 |
BC018141.1 | - | 2 - 3 | 1623 | 64 | |
CD014089.1 | - | 2 - 3 | 1673 | 64 | |
M16474.1 | - | 2 - 3 | 1591 | 64 | |
M16541.1 | - | 2 - 3 | 1645 | 64 |