Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CYP2D6
- Entrez Gene ID
-
1565
- Gene Name
- cytochrome P450 family 2 subfamily D member 6
- Gene Aliases
-
CPD6, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, P450-DB1, P450C2D, P450DB1
- Chromosome Location
-
Chr.22: 42125531 - 42130881 on Build GRCh38
- Gene Symbol
- LOC101929829
- Entrez Gene ID
-
101929829
- Gene Name
- cytochrome P450 family 2 subfamily D member 6 pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_111920.1 |
|
2 - 3 |
445 |
74 |
GenBank mRNA |
AB209492.1 |
- |
1 - 2 |
2624 |
74 |
AK308211.1 |
- |
2 - 3 |
440 |
74 |
DA034539.1 |
- |
2 - 3 |
445 |
74 |
View Full Product Details