Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GBA
- Entrez Gene ID
-
2629
- Gene Name
- glucosylceramidase beta
- Gene Aliases
-
GBA1, GCB, GLUC
- Chromosome Location
-
Chr.1: 155234448 - 155244862 on Build GRCh38
- Gene Symbol
- GBAP1
- Entrez Gene ID
-
2630
- Gene Name
- glucosylceramidase beta pseudogene 1
- Gene Aliases
-
GBAP
- Chromosome Location
-
Chr.1: 155213825 - 155227534 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
KJ690774.1 |
- |
8 - 9 |
1305 |
85 |
KJ690776.1 |
- |
9 - 10 |
1195 |
85 |
View Full Product Details