The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000423.2 | NP_000414.2 | 4 - 5 | 991 | 98 |
GenBank mRNA | BC096294.4 | - | 4 - 5 | 980 | 98 |
BC099643.1 | - | 4 - 5 | 980 | 98 | |
BC099644.3 | - | 4 - 5 | 980 | 98 | |
M99061.1 | - | 4 - 5 | 1009 | 98 |