The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000555.3 | NP_000546.2 | 4 - 5 | 1227 | 77 |
NM_001195553.1 | NP_001182482.1 | 4 - 5 | 929 | 77 | |
NM_178151.2 | NP_835364.1 | 4 - 5 | 1056 | 77 | |
NM_178152.2 | NP_835365.1 | 4 - 5 | 929 | 77 | |
NM_178153.2 | NP_835366.1 | 4 - 5 | 929 | 77 | |
XM_011530878.2 | XP_011529180.1 | 4 - 5 | 901 | 77 | |
XM_011530879.2 | XP_011529181.1 | 4 - 5 | 901 | 77 | |
XM_011530880.2 | XP_011529182.1 | 4 - 5 | 901 | 77 | |
XM_017029312.1 | XP_016884801.1 | 4 - 5 | 994 | 77 | |
GenBank mRNA | AF034634.1 | - | 3 - 4 | 812 | 77 |
AF040254.1 | - | 4 - 5 | 854 | 77 | |
AF040255.1 | - | 4 - 5 | 854 | 77 | |
AJ003112.1 | - | 4 - 5 | 1227 | 77 | |
AK290455.1 | - | 4 - 5 | 876 | 77 | |
AK297297.1 | - | 4 - 5 | 1030 | 77 | |
BC027925.1 | - | 4 - 5 | 871 | 77 |