Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60444625-60445171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APPBP2
Entrez Gene ID
10513
Gene Name
amyloid beta precursor protein binding protein 2
Gene Aliases
APP-BP2, HS.84084, PAT1
UniGene
Hs.84084
Gene Chromosome Location
Chr.17: 60443149-60526240 on Build GRCh38
Transcripts
NM_001282476.1 NM_006380.3 XM_011524198.2 XM_011524199.2

SNP Detail

SNP ID Chromosome Location
rs912211585 Chr.17: 60444723-60444723 on Build GRCh38
rs1010185171 Chr.17: 60444823-60444823 on Build GRCh38
rs938626080 Chr.17: 60445039-60445039 on Build GRCh38
rs192683170 Chr.17: 60444693-60444693 on Build GRCh38
rs969190917 Chr.17: 60444833-60444833 on Build GRCh38
rs954718352 Chr.17: 60444819-60444819 on Build GRCh38
rs996326301 Chr.17: 60444971-60444971 on Build GRCh38
rs948991682 Chr.17: 60444707-60444707 on Build GRCh38
rs370391168 Chr.17: 60445079-60445080 on Build GRCh38
rs1030515787 Chr.17: 60444771-60444771 on Build GRCh38
rs1052000990 Chr.17: 60444987-60444987 on Build GRCh38
rs890705625 Chr.17: 60445008-60445008 on Build GRCh38
rs973911526 Chr.17: 60444673-60444673 on Build GRCh38
rs532442905 Chr.17: 60445076-60445077 on Build GRCh38
rs1055021447 Chr.17: 60444684-60444684 on Build GRCh38
rs771879362 Chr.17: 60444735-60444735 on Build GRCh38
rs893326681 Chr.17: 60444761-60444761 on Build GRCh38
rs764767630 Chr.17: 60444933-60444933 on Build GRCh38
rs183222206 Chr.17: 60444736-60444736 on Build GRCh38
rs151181942 Chr.17: 60445095-60445095 on Build GRCh38
rs1048359986 Chr.17: 60445117-60445117 on Build GRCh38
rs532818785 Chr.17: 60444948-60444948 on Build GRCh38
rs1037426216 Chr.17: 60444950-60444950 on Build GRCh38
rs770058878 Chr.17: 60445094-60445094 on Build GRCh38
rs142914675 Chr.17: 60444829-60444829 on Build GRCh38
rs547203871 Chr.17: 60444879-60444879 on Build GRCh38
rs16944359 Chr.17: 60445134-60445134 on Build GRCh38
rs995462341 Chr.17: 60444839-60444839 on Build GRCh38
rs914730254 Chr.17: 60444955-60444955 on Build GRCh38
rs896174657 Chr.17: 60444690-60444690 on Build GRCh38
rs990740788 Chr.17: 60444870-60444870 on Build GRCh38
rs1047424505 Chr.17: 60444880-60444880 on Build GRCh38
rs746943074 Chr.17: 60444963-60444963 on Build GRCh38
rs763892055 Chr.17: 60444844-60444844 on Build GRCh38
rs1047518196 Chr.17: 60444708-60444708 on Build GRCh38
rs559854949 Chr.17: 60444751-60444751 on Build GRCh38
rs571187153 Chr.17: 60445000-60445000 on Build GRCh38
rs193042374 Chr.17: 60444832-60444832 on Build GRCh38
rs750138239 Chr.17: 60445069-60445069 on Build GRCh38
rs976275927 Chr.17: 60444739-60444739 on Build GRCh38
rs761488296 Chr.17: 60444922-60444922 on Build GRCh38
rs188418340 Chr.17: 60444760-60444760 on Build GRCh38
rs184867340 Chr.17: 60444856-60444856 on Build GRCh38
rs943720016 Chr.17: 60444729-60444729 on Build GRCh38
rs956280937 Chr.17: 60444941-60444941 on Build GRCh38
rs189965326 Chr.17: 60444952-60444952 on Build GRCh38
rs996844347 Chr.17: 60444770-60444770 on Build GRCh38
rs1015486661 Chr.17: 60444813-60444813 on Build GRCh38
rs1026805973 Chr.17: 60444843-60444843 on Build GRCh38
rs866348106 Chr.17: 60444794-60444794 on Build GRCh38
rs776313287 Chr.17: 60444854-60444854 on Build GRCh38
rs1044458608 Chr.17: 60445022-60445022 on Build GRCh38
rs747918900 Chr.17: 60444686-60444686 on Build GRCh38
rs58325641 Chr.17: 60444947-60444947 on Build GRCh38
rs977728869 Chr.17: 60444924-60444928 on Build GRCh38
rs928720647 Chr.17: 60445030-60445030 on Build GRCh38
rs907179364 Chr.17: 60444805-60444805 on Build GRCh38
rs9898327 Chr.17: 60444677-60444677 on Build GRCh38
rs1044899837 Chr.17: 60444787-60444787 on Build GRCh38
rs917804075 Chr.17: 60444874-60444874 on Build GRCh38
rs1038255724 Chr.17: 60444752-60444752 on Build GRCh38

View Full Product Details