Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60451890-60452439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APPBP2
Entrez Gene ID
10513
Gene Name
amyloid beta precursor protein binding protein 2
Gene Aliases
APP-BP2, HS.84084, PAT1
UniGene
Hs.84084
Gene Chromosome Location
Chr.17: 60443149-60526240 on Build GRCh38
Transcripts
NM_001282476.1 NM_006380.3 XM_011524198.2 XM_011524199.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3362262 c.1382A>C p.Q461P Chr.17: 60452002-60452002 on Build GRCh38
6423538 c.1411G>T p.A471S Chr.17: 60451973-60451973 on Build GRCh38
6189961 c.1378G>A p.E460K Chr.17: 60452006-60452006 on Build GRCh38
5804626 c.1347A>T p.E449D Chr.17: 60452037-60452037 on Build GRCh38
6260880 c.1489C>A p.R497R Chr.17: 60451895-60451895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906105025 Chr.17: 60452333-60452333 on Build GRCh38
rs947875169 Chr.17: 60452214-60452214 on Build GRCh38
rs774163000 Chr.17: 60451931-60451931 on Build GRCh38
rs182053463 Chr.17: 60452195-60452195 on Build GRCh38
rs7212360 Chr.17: 60452040-60452040 on Build GRCh38
rs530334686 Chr.17: 60452178-60452178 on Build GRCh38
rs906426351 Chr.17: 60452225-60452225 on Build GRCh38
rs550370704 Chr.17: 60452242-60452242 on Build GRCh38
rs560617765 Chr.17: 60452202-60452202 on Build GRCh38
rs771471911 Chr.17: 60451995-60451995 on Build GRCh38
rs528128234 Chr.17: 60452209-60452209 on Build GRCh38
rs1028689725 Chr.17: 60452370-60452370 on Build GRCh38
rs750877479 Chr.17: 60452069-60452069 on Build GRCh38
rs187158361 Chr.17: 60452211-60452211 on Build GRCh38
rs749018643 Chr.17: 60452092-60452092 on Build GRCh38
rs7212665 Chr.17: 60452213-60452213 on Build GRCh38
rs755148113 Chr.17: 60452075-60452075 on Build GRCh38
rs376089888 Chr.17: 60452074-60452074 on Build GRCh38
rs781410508 Chr.17: 60452077-60452077 on Build GRCh38
rs1057279903 Chr.17: 60452381-60452381 on Build GRCh38
rs755093801 Chr.17: 60452097-60452097 on Build GRCh38
rs762270239 Chr.17: 60452062-60452062 on Build GRCh38
rs759587878 Chr.17: 60451971-60451971 on Build GRCh38
rs946091593 Chr.17: 60452108-60452108 on Build GRCh38
rs536057101 Chr.17: 60452322-60452322 on Build GRCh38
rs775115797 Chr.17: 60452003-60452003 on Build GRCh38
rs977406104 Chr.17: 60452159-60452159 on Build GRCh38
rs536714840 Chr.17: 60452222-60452222 on Build GRCh38
rs895988670 Chr.17: 60452408-60452408 on Build GRCh38
rs1014561194 Chr.17: 60452288-60452288 on Build GRCh38
rs760213946 Chr.17: 60452016-60452016 on Build GRCh38
rs1025619004 Chr.17: 60452155-60452155 on Build GRCh38
rs947596689 Chr.17: 60451965-60451965 on Build GRCh38
rs373202345 Chr.17: 60452060-60452060 on Build GRCh38
rs765762179 Chr.17: 60452066-60452066 on Build GRCh38
rs983967759 Chr.17: 60452174-60452174 on Build GRCh38
rs988316567 Chr.17: 60452248-60452248 on Build GRCh38
rs994681682 Chr.17: 60452112-60452112 on Build GRCh38
rs925914537 Chr.17: 60452162-60452162 on Build GRCh38
rs1046350978 Chr.17: 60452217-60452217 on Build GRCh38
rs1004535236 Chr.17: 60452229-60452229 on Build GRCh38
rs188883909 Chr.17: 60452276-60452276 on Build GRCh38
rs898793644 Chr.17: 60452308-60452308 on Build GRCh38
rs770801236 Chr.17: 60451921-60451921 on Build GRCh38
rs912128015 Chr.17: 60452089-60452089 on Build GRCh38
rs758990402 Chr.17: 60452073-60452073 on Build GRCh38
rs561431317 Chr.17: 60452110-60452110 on Build GRCh38

View Full Product Details