The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000689.4 | NP_000680.2 | 11 - 12 | 1675 | 97 |
GenBank mRNA | AB209821.1 | - | 11 - 12 | 1402 | 97 |
AF003341.1 | - | 11 - 12 | 1358 | 97 | |
AK000118.1 | - | 11 - 12 | 1402 | 97 | |
AK026641.1 | - | 11 - 12 | 1402 | 97 | |
AK293176.1 | - | 11 - 12 | 1368 | 97 | |
AY390731.1 | - | 11 - 12 | 1358 | 97 | |
BC001505.2 | - | 11 - 12 | 1383 | 97 | |
BT006921.1 | - | 11 - 12 | 1358 | 97 | |
EU668318.1 | - | 11 - 12 | 1402 | 97 | |
EU794589.1 | - | 11 - 12 | 1358 | 97 | |
FJ224286.1 | - | 11 - 12 | 1411 | 97 | |
K03000.1 | - | 7 - 8 | 875 | 97 | |
M26761.1 | - | 7 - 8 | 875 | 97 |