Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45396115-45396605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP27
Entrez Gene ID
201176
Gene Name
Rho GTPase activating protein 27
Gene Aliases
CAMGAP1, PP905, SH3D20, SH3P20
UniGene
Hs.205326
Gene Chromosome Location
Chr.17: 45393902-45432916 on Build GRCh38
Transcripts
NM_001282290.1 NM_199282.2 XM_005257128.3 XM_006721745.2 XM_006721747.2 XM_011524468.1 XM_011524469.1 XM_011524470.2 XM_011524471.1 XM_011524472.2 XM_011524474.1 XM_017024311.1 XM_017024312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4753901 c.1174C>T p.R392C Chr.17: 45396261-45396261 on Build GRCh38
6362009 c.2095C>A p.L699M Chr.17: 45396565-45396565 on Build GRCh38
6362008 c.1072C>A p.L358M Chr.17: 45396565-45396565 on Build GRCh38
6081154 c.2135G>T p.R712L Chr.17: 45396525-45396525 on Build GRCh38
6081153 c.1112G>T p.R371L Chr.17: 45396525-45396525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778940620 Chr.17: 45396323-45396324 on Build GRCh38
rs775059872 Chr.17: 45396218-45396218 on Build GRCh38
rs753830103 Chr.17: 45396231-45396231 on Build GRCh38
rs758150391 Chr.17: 45396246-45396246 on Build GRCh38
rs761140543 Chr.17: 45396311-45396311 on Build GRCh38
rs369152481 Chr.17: 45396575-45396575 on Build GRCh38
rs201648544 Chr.17: 45396207-45396207 on Build GRCh38
rs766398685 Chr.17: 45396149-45396149 on Build GRCh38
rs146946789 Chr.17: 45396324-45396324 on Build GRCh38
rs530491453 Chr.17: 45396271-45396271 on Build GRCh38
rs375387387 Chr.17: 45396578-45396578 on Build GRCh38
rs113604190 Chr.17: 45396273-45396273 on Build GRCh38
rs893851153 Chr.17: 45396450-45396450 on Build GRCh38
rs201032827 Chr.17: 45396467-45396467 on Build GRCh38
rs754847357 Chr.17: 45396156-45396156 on Build GRCh38
rs34337698 Chr.17: 45396387-45396388 on Build GRCh38
rs905632681 Chr.17: 45396373-45396373 on Build GRCh38
rs764060794 Chr.17: 45396452-45396452 on Build GRCh38
rs765145988 Chr.17: 45396235-45396235 on Build GRCh38
rs374691739 Chr.17: 45396201-45396201 on Build GRCh38
rs868490141 Chr.17: 45396443-45396443 on Build GRCh38
rs368477600 Chr.17: 45396179-45396179 on Build GRCh38
rs755803083 Chr.17: 45396559-45396559 on Build GRCh38
rs748692786 Chr.17: 45396294-45396294 on Build GRCh38
rs775518547 Chr.17: 45396544-45396544 on Build GRCh38
rs745427174 Chr.17: 45396204-45396204 on Build GRCh38
rs759726287 Chr.17: 45396232-45396232 on Build GRCh38
rs748854028 Chr.17: 45396505-45396505 on Build GRCh38
rs750099267 Chr.17: 45396490-45396490 on Build GRCh38
rs771984863 Chr.17: 45396483-45396492 on Build GRCh38
rs774362215 Chr.17: 45396552-45396552 on Build GRCh38
rs769446965 Chr.17: 45396276-45396276 on Build GRCh38
rs371609350 Chr.17: 45396197-45396197 on Build GRCh38
rs745359488 Chr.17: 45396275-45396275 on Build GRCh38
rs778408571 Chr.17: 45396513-45396513 on Build GRCh38
rs767380929 Chr.17: 45396557-45396557 on Build GRCh38
rs920839951 Chr.17: 45396558-45396558 on Build GRCh38
rs947792766 Chr.17: 45396387-45396387 on Build GRCh38
rs746015840 Chr.17: 45396469-45396469 on Build GRCh38
rs533078667 Chr.17: 45396568-45396568 on Build GRCh38
rs771339682 Chr.17: 45396521-45396521 on Build GRCh38
rs760827805 Chr.17: 45396224-45396224 on Build GRCh38
rs144020235 Chr.17: 45396221-45396221 on Build GRCh38
rs1018261139 Chr.17: 45396462-45396462 on Build GRCh38
rs922677598 Chr.17: 45396471-45396471 on Build GRCh38
rs777341708 Chr.17: 45396190-45396190 on Build GRCh38
rs562893433 Chr.17: 45396532-45396532 on Build GRCh38
rs745671602 Chr.17: 45396429-45396429 on Build GRCh38
rs544328670 Chr.17: 45396528-45396528 on Build GRCh38
rs763927588 Chr.17: 45396548-45396548 on Build GRCh38
rs377722433 Chr.17: 45396166-45396166 on Build GRCh38
rs758940297 Chr.17: 45396508-45396508 on Build GRCh38
rs540086629 Chr.17: 45396404-45396404 on Build GRCh38
rs770992376 Chr.17: 45396143-45396143 on Build GRCh38
rs769677156 Chr.17: 45396483-45396483 on Build GRCh38
rs988051659 Chr.17: 45396160-45396160 on Build GRCh38
rs371679665 Chr.17: 45396502-45396502 on Build GRCh38
rs758246377 Chr.17: 45396183-45396183 on Build GRCh38
rs926921928 Chr.17: 45396537-45396537 on Build GRCh38
rs763089890 Chr.17: 45396545-45396545 on Build GRCh38
rs754592200 Chr.17: 45396571-45396571 on Build GRCh38
rs374182883 Chr.17: 45396239-45396239 on Build GRCh38
rs750691619 Chr.17: 45396142-45396142 on Build GRCh38
rs779791561 Chr.17: 45396534-45396534 on Build GRCh38
rs367713495 Chr.17: 45396330-45396330 on Build GRCh38
rs778083025 Chr.17: 45396574-45396574 on Build GRCh38
rs766669825 Chr.17: 45396225-45396225 on Build GRCh38
rs937004858 Chr.17: 45396551-45396551 on Build GRCh38
rs762214702 Chr.17: 45396219-45396219 on Build GRCh38
rs772512075 Chr.17: 45396296-45396296 on Build GRCh38
rs372585905 Chr.17: 45396342-45396342 on Build GRCh38
rs760737219 Chr.17: 45396145-45396145 on Build GRCh38
rs555932507 Chr.17: 45396488-45396488 on Build GRCh38
rs368506644 Chr.17: 45396335-45396335 on Build GRCh38
rs776969420 Chr.17: 45396328-45396328 on Build GRCh38
rs747320660 Chr.17: 45396520-45396520 on Build GRCh38
rs146454541 Chr.17: 45396237-45396237 on Build GRCh38
rs762856719 Chr.17: 45396442-45396442 on Build GRCh38
rs575887643 Chr.17: 45396344-45396344 on Build GRCh38
rs779499425 Chr.17: 45396278-45396278 on Build GRCh38
rs779728849 Chr.17: 45396504-45396504 on Build GRCh38
rs999470353 Chr.17: 45396418-45396418 on Build GRCh38
rs540748930 Chr.17: 45396474-45396474 on Build GRCh38
rs1046676745 Chr.17: 45396399-45396399 on Build GRCh38
rs751421164 Chr.17: 45396264-45396264 on Build GRCh38
rs761451878 Chr.17: 45396556-45396556 on Build GRCh38
rs558073474 Chr.17: 45396423-45396423 on Build GRCh38
rs752484395 Chr.17: 45396175-45396175 on Build GRCh38
rs777568132 Chr.17: 45396247-45396247 on Build GRCh38
rs751310963 Chr.17: 45396461-45396461 on Build GRCh38
rs984197223 Chr.17: 45396582-45396582 on Build GRCh38
rs773423248 Chr.17: 45396298-45396298 on Build GRCh38
rs374952036 Chr.17: 45396155-45396155 on Build GRCh38
rs777985349 Chr.17: 45396376-45396376 on Build GRCh38
rs972329056 Chr.17: 45396526-45396526 on Build GRCh38
rs765332100 Chr.17: 45396333-45396333 on Build GRCh38
rs147589670 Chr.17: 45396569-45396569 on Build GRCh38
rs746672599 Chr.17: 45396195-45396195 on Build GRCh38
rs770051091 Chr.17: 45396543-45396543 on Build GRCh38
rs17630836 Chr.17: 45396274-45396274 on Build GRCh38
rs778859341 Chr.17: 45396169-45396169 on Build GRCh38
rs983677075 Chr.17: 45396477-45396477 on Build GRCh38
rs772565283 Chr.17: 45396220-45396220 on Build GRCh38
rs987545972 Chr.17: 45396499-45396499 on Build GRCh38

View Full Product Details