The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000733.3 | NP_000724.1 | 2 - 3 | 307 | 89 |
GenBank mRNA | AK292612.1 | - | 2 - 3 | 156 | 89 |
AK311641.1 | - | 2 - 3 | 157 | 89 | |
BI910359.1 | - | 2 - 3 | 342 | 89 | |
BP366535.1 | - | 2 - 3 | 324 | 89 | |
X03884.1 | - | 1 - 2 | 105 | 89 |