Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRM6
Entrez Gene ID
2916
Gene Name
glutamate metabotropic receptor 6
Gene Aliases
CSNB1B, GPRC1F, MGLUR6, mGlu6
Chromosome Location
Chr.5: 178978327 - 178995123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ245871.1 - 2 - 3 123 115
AJ245872.1 - 2 - 3 225 115

View Full Product Details