Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRIM2
Entrez Gene ID
5558
Gene Name
primase (DNA) subunit 2
Gene Aliases
PRIM2A, p58
Chromosome Location
Chr.6: 57314805 - 57646852 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000947.4 NP_000938.2 9 - 10 923 71
XM_011514731.2 XP_011513033.1 3 - 4 2690 71
XM_017011004.1 XP_016866493.1 10 - 11 1115 71
XM_017011005.1 XP_016866494.1 10 - 11 1115 71
XM_017011006.1 XP_016866495.1 10 - 11 1115 71
XM_017011007.1 XP_016866496.1 7 - 8 770 71
GenBank mRNA AK291915.1 - 9 - 10 910 71
BC064931.1 - 9 - 10 908 71
X74331.1 - 9 - 10 916 71

View Full Product Details