Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50699686-50700166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD40
Entrez Gene ID
91369
Gene Name
ankyrin repeat domain 40
Gene Aliases
-
UniGene
Hs.463426
Gene Chromosome Location
Chr.17: 50693190-50707909 on Build GRCh38
Transcripts
NM_052855.3 XM_017025296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
334063 c.469G>T p.A157S Chr.17: 50699708-50699708 on Build GRCh38
7181873 c.333G>T p.L111L Chr.17: 50699844-50699844 on Build GRCh38
7174161 c.429G>A p.M143I Chr.17: 50699748-50699748 on Build GRCh38
7110064 c.312_317delTGATGA p.D105_D106delDD Chr.17: 50699860-50699865 on Build GRCh38
706886 c.443C>T p.P148L Chr.17: 50699734-50699734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968225274 Chr.17: 50699867-50699867 on Build GRCh38
rs983213568 Chr.17: 50700078-50700078 on Build GRCh38
rs78012840 Chr.17: 50700077-50700077 on Build GRCh38
rs918938161 Chr.17: 50699929-50699929 on Build GRCh38
rs145395652 Chr.17: 50699880-50699880 on Build GRCh38
rs752303583 Chr.17: 50699834-50699834 on Build GRCh38
rs774155277 Chr.17: 50699860-50699868 on Build GRCh38
rs138572332 Chr.17: 50699859-50699860 on Build GRCh38
rs911685961 Chr.17: 50699885-50699885 on Build GRCh38
rs757230507 Chr.17: 50699865-50699865 on Build GRCh38
rs569194211 Chr.17: 50700062-50700062 on Build GRCh38
rs202219117 Chr.17: 50699757-50699757 on Build GRCh38
rs778789385 Chr.17: 50699870-50699870 on Build GRCh38
rs77108941 Chr.17: 50699761-50699761 on Build GRCh38
rs776919239 Chr.17: 50699927-50699927 on Build GRCh38
rs755909727 Chr.17: 50699860-50699871 on Build GRCh38
rs753650876 Chr.17: 50699861-50699861 on Build GRCh38
rs199734034 Chr.17: 50699883-50699883 on Build GRCh38
rs748907253 Chr.17: 50699881-50699883 on Build GRCh38
rs761935069 Chr.17: 50699986-50699986 on Build GRCh38
rs770725227 Chr.17: 50699880-50699881 on Build GRCh38
rs944457398 Chr.17: 50699894-50699894 on Build GRCh38
rs755724767 Chr.17: 50699842-50699842 on Build GRCh38
rs200713460 Chr.17: 50699852-50699852 on Build GRCh38
rs745922334 Chr.17: 50699726-50699726 on Build GRCh38
rs896162937 Chr.17: 50699838-50699840 on Build GRCh38
rs150044915 Chr.17: 50699850-50699850 on Build GRCh38
rs760904813 Chr.17: 50699758-50699758 on Build GRCh38
rs915053281 Chr.17: 50699779-50699779 on Build GRCh38
rs767002870 Chr.17: 50699830-50699830 on Build GRCh38
rs1056269266 Chr.17: 50699828-50699828 on Build GRCh38
rs751166430 Chr.17: 50699790-50699790 on Build GRCh38
rs755882699 Chr.17: 50699711-50699711 on Build GRCh38
rs1049325694 Chr.17: 50700117-50700117 on Build GRCh38
rs778505031 Chr.17: 50699893-50699894 on Build GRCh38
rs770465484 Chr.17: 50699938-50699938 on Build GRCh38
rs765827384 Chr.17: 50699776-50699776 on Build GRCh38
rs975670113 Chr.17: 50699959-50699959 on Build GRCh38
rs774469514 Chr.17: 50699722-50699722 on Build GRCh38
rs767987488 Chr.17: 50700138-50700138 on Build GRCh38
rs774926022 Chr.17: 50699860-50699862 on Build GRCh38
rs955607873 Chr.17: 50700051-50700051 on Build GRCh38
rs149209063 Chr.17: 50699893-50699893 on Build GRCh38
rs776991784 Chr.17: 50699763-50699763 on Build GRCh38
rs762245683 Chr.17: 50699931-50699931 on Build GRCh38
rs759087532 Chr.17: 50699829-50699829 on Build GRCh38
rs922885517 Chr.17: 50700025-50700025 on Build GRCh38
rs865806977 Chr.17: 50699767-50699767 on Build GRCh38
rs770850055 Chr.17: 50699718-50699718 on Build GRCh38
rs762478370 Chr.17: 50699768-50699768 on Build GRCh38
rs774406645 Chr.17: 50699923-50699923 on Build GRCh38
rs772164531 Chr.17: 50699882-50699882 on Build GRCh38
rs1002092660 Chr.17: 50699723-50699723 on Build GRCh38
rs745442245 Chr.17: 50699897-50699901 on Build GRCh38
rs564401651 Chr.17: 50699716-50699716 on Build GRCh38
rs930876531 Chr.17: 50700083-50700083 on Build GRCh38
rs750724196 Chr.17: 50699862-50699863 on Build GRCh38
rs539996292 Chr.17: 50699712-50699712 on Build GRCh38

View Full Product Details