Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50707391-50707930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD40
Entrez Gene ID
91369
Gene Name
ankyrin repeat domain 40
Gene Aliases
-
UniGene
Hs.463426
Gene Chromosome Location
Chr.17: 50693190-50707909 on Build GRCh38
Transcripts
NM_052855.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6199984 c.79C>T p.Q27* Chr.17: 50707576-50707576 on Build GRCh38
3192925 c.51C>G p.A17A Chr.17: 50707604-50707604 on Build GRCh38
4652872 c.117A>G p.Q39Q Chr.17: 50707538-50707538 on Build GRCh38
3192924 c.60A>G p.L20L Chr.17: 50707595-50707595 on Build GRCh38
6600724 c.7G>A p.A3T Chr.17: 50707648-50707648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535785206 Chr.17: 50707752-50707752 on Build GRCh38
rs754804181 Chr.17: 50707579-50707579 on Build GRCh38
rs147758912 Chr.17: 50707623-50707623 on Build GRCh38
rs754100699 Chr.17: 50707692-50707692 on Build GRCh38
rs551868482 Chr.17: 50707584-50707584 on Build GRCh38
rs766319929 Chr.17: 50707476-50707476 on Build GRCh38
rs117204968 Chr.17: 50707573-50707573 on Build GRCh38
rs762777651 Chr.17: 50707556-50707556 on Build GRCh38
rs774011618 Chr.17: 50707675-50707675 on Build GRCh38
rs917599411 Chr.17: 50707611-50707611 on Build GRCh38
rs932853863 Chr.17: 50707866-50707866 on Build GRCh38
rs199621660 Chr.17: 50707509-50707509 on Build GRCh38
rs921693310 Chr.17: 50707852-50707852 on Build GRCh38
rs769477941 Chr.17: 50707649-50707649 on Build GRCh38
rs1012881116 Chr.17: 50707488-50707488 on Build GRCh38
rs374500997 Chr.17: 50707506-50707506 on Build GRCh38
rs770775422 Chr.17: 50707670-50707670 on Build GRCh38
rs749426972 Chr.17: 50707617-50707617 on Build GRCh38
rs894476724 Chr.17: 50707442-50707442 on Build GRCh38
rs764860815 Chr.17: 50707538-50707538 on Build GRCh38
rs752864828 Chr.17: 50707683-50707683 on Build GRCh38
rs529233671 Chr.17: 50707467-50707467 on Build GRCh38
rs200569179 Chr.17: 50707669-50707669 on Build GRCh38
rs772566107 Chr.17: 50707645-50707645 on Build GRCh38
rs3833149 Chr.17: 50707684-50707684 on Build GRCh38
rs756399469 Chr.17: 50707490-50707490 on Build GRCh38
rs550801978 Chr.17: 50707646-50707646 on Build GRCh38
rs1015537672 Chr.17: 50707690-50707691 on Build GRCh38
rs997114746 Chr.17: 50707676-50707676 on Build GRCh38
rs773589613 Chr.17: 50707678-50707679 on Build GRCh38
rs749647375 Chr.17: 50707507-50707507 on Build GRCh38
rs555537335 Chr.17: 50707758-50707758 on Build GRCh38
rs3744519 Chr.17: 50707739-50707739 on Build GRCh38
rs746309044 Chr.17: 50707512-50707512 on Build GRCh38
rs751630695 Chr.17: 50707478-50707478 on Build GRCh38
rs962783271 Chr.17: 50707720-50707720 on Build GRCh38
rs936405709 Chr.17: 50707834-50707834 on Build GRCh38
rs764217338 Chr.17: 50707687-50707687 on Build GRCh38
rs757351749 Chr.17: 50707694-50707694 on Build GRCh38
rs770016206 Chr.17: 50707665-50707666 on Build GRCh38
rs991999098 Chr.17: 50707789-50707789 on Build GRCh38
rs758956973 Chr.17: 50707701-50707701 on Build GRCh38
rs757661096 Chr.17: 50707621-50707621 on Build GRCh38
rs899958213 Chr.17: 50707671-50707672 on Build GRCh38
rs779403002 Chr.17: 50707695-50707695 on Build GRCh38
rs8068442 Chr.17: 50707804-50707804 on Build GRCh38
rs767770817 Chr.17: 50707585-50707585 on Build GRCh38
rs539546410 Chr.17: 50707519-50707519 on Build GRCh38
rs756452773 Chr.17: 50707601-50707601 on Build GRCh38
rs750742504 Chr.17: 50707700-50707700 on Build GRCh38
rs767539915 Chr.17: 50707681-50707681 on Build GRCh38
rs760865098 Chr.17: 50707684-50707684 on Build GRCh38
rs766049572 Chr.17: 50707565-50707565 on Build GRCh38
rs778106424 Chr.17: 50707605-50707605 on Build GRCh38
rs748384276 Chr.17: 50707487-50707487 on Build GRCh38
rs560471352 Chr.17: 50707451-50707451 on Build GRCh38
rs974862200 Chr.17: 50707841-50707841 on Build GRCh38
rs752810096 Chr.17: 50707588-50707588 on Build GRCh38
rs544255374 Chr.17: 50707808-50707808 on Build GRCh38
rs959434304 Chr.17: 50707774-50707774 on Build GRCh38
rs1042056387 Chr.17: 50707642-50707642 on Build GRCh38
rs755119066 Chr.17: 50707479-50707479 on Build GRCh38
rs761522635 Chr.17: 50707532-50707532 on Build GRCh38
rs940249788 Chr.17: 50707909-50707909 on Build GRCh38
rs146954333 Chr.17: 50707651-50707651 on Build GRCh38
rs12601696 Chr.17: 50707678-50707678 on Build GRCh38
rs747638860 Chr.17: 50707647-50707647 on Build GRCh38
rs546840335 Chr.17: 50707438-50707438 on Build GRCh38
rs773043508 Chr.17: 50707544-50707544 on Build GRCh38
rs891409349 Chr.17: 50707908-50707908 on Build GRCh38
rs1024920011 Chr.17: 50707803-50707803 on Build GRCh38
rs1051256776 Chr.17: 50707906-50707913 on Build GRCh38
rs1049644165 Chr.17: 50707410-50707410 on Build GRCh38
rs781374570 Chr.17: 50707483-50707483 on Build GRCh38
rs925052519 Chr.17: 50707643-50707643 on Build GRCh38
rs879122681 Chr.17: 50707733-50707733 on Build GRCh38

View Full Product Details