The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000591.3 | NP_000582.1 | 1 - 2 | 383 | 63 |
NM_001040021.2 | NP_001035110.1 | 2 - 3 | 296 | 63 | |
NM_001174104.1 | NP_001167575.1 | 2 - 3 | 396 | 63 | |
NM_001174105.1 | NP_001167576.1 | 2 - 3 | 275 | 63 | |
GenBank mRNA | AK313277.1 | - | 1 - 2 | 108 | 63 |
BC010507.2 | - | 1 - 2 | 109 | 63 | |
BP300412.1 | - | 2 - 3 | 395 | 63 | |
CF593452.1 | - | 1 - 2 | 634 | 63 | |
DA269805.1 | - | 2 - 3 | 251 | 63 | |
DA426297.1 | - | 2 - 3 | 296 | 63 | |
DA681405.1 | - | 2 - 3 | 265 | 63 | |
M86511.1 | - | 1 - 2 | 79 | 63 | |
X13334.1 | - | 1 - 2 | 123 | 63 |