The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002583.2 | NP_002574.2 | 3 - 4 | 936 | 126 |
XM_006719435.2 | XP_006719498.1 | 3 - 4 | 912 | 126 | |
XM_017019377.1 | XP_016874866.1 | 3 - 4 | 816 | 126 | |
GenBank mRNA | BC007018.1 | - | 2 - 3 | 782 | 126 |
CR536549.1 | - | 2 - 3 | 650 | 126 | |
U63809.1 | - | 3 - 4 | 894 | 126 |