The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016640.3 | NP_057724.2 | 1 - 2 | 637 | 85 |
GenBank mRNA | AF146192.2 | - | 1 - 2 | 637 | 85 |
AF217523.1 | - | 1 - 2 | 83 | ||
AK074777.1 | - | 1 - 2 | 605 | 85 | |
AK222704.1 | - | 1 - 2 | 615 | 85 | |
AK222894.1 | - | 1 - 2 | 607 | 85 | |
AL136706.1 | - | 1 - 2 | 610 | 85 | |
AL355715.1 | - | 1 - 2 | 637 | 85 | |
AL355716.1 | - | 1 - 2 | 330 | 85 | |
BC002460.2 | - | 1 - 2 | 588 | 85 | |
BC007735.2 | - | 1 - 2 | 614 | 85 |