Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INSR
Entrez Gene ID
3643
Gene Name
insulin receptor
Gene Aliases
CD220, HHF5
Chromosome Location
Chr.19: 7112255 - 7294405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000208.3 NP_000199.2 11 - 12 2682 90
XM_011527988.2 XP_011526290.2 11 - 12 2785 90
GenBank mRNA M10051.1 - 11 - 12 2409 90
M24555.1 - 2 - 3 90 90

View Full Product Details