The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001264.4 | NP_001255.3 | 1 - 2 | 145 | 79 |
GenBank mRNA | AF030130.1 | - | 1 - 2 | 103 | 79 |
AU134804.1 | - | 1 - 2 | 117 | 79 |